Schlenk Pear Shaped Storage Flask with Sidearm and Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Used for storage, but may also be used for simple reactions
With standard taper 14/20 or standard taper 24/40 inner joint or #15 o-ring joint and 2 or 4 mm bore glass stopcock or 2 mm inlet valve. Fabricated from borosilicate glass.
Frequently Bought Together