HPLC carts contribute to greater benchtop productivity.
- Mobilize and stack HPLC equipment on carts to maximize valuable lab space
- Avoid crowding valuable bench top space with HPLCor other analytical equipment
Five-shelf units are mounted on 188cm (74") posts with casters and are used for stacking and mobilizing HPLC or other analytical equipment
Slide-out keyboard shelf is epoxy-coated for a high level of corrosion resistance. Flat monitor mounting kit features a unique bracket and crossbar design for mounting flat computer monitors between two MetroMax i posts, either in back or front of unit. Six-outlet power strip with surge protection mounts flush with the shelf frame and includes a 1.8m (6') cord. Power strip is UL listed.
They feature strong polymer construction with superior corrosion resistance. Shelves are adjustable in 2.5cm (1") increments along the height of the post. Each five-shelf cart has four solid shelves and one open-grid shelf. Each shelf includes a mat made from mineral-reinforced polypropylene with built-in antimicrobial protection from bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Carts can be used in cold conditions as low as –29°C (–20°F).