Semi-Volatile Surrogate Standards for EPA methods, Restek
RK31091
: Restek
:
- Description:2-Fluorobiphenyl Standard
- Cat. no.:RK31091
- Supplier No.:31091
Standards are packaged in methylene chloride at a concentration of 2000 µg/mL in a 1 mL ampoule.
- Mixture contains 2-Fluorobiphenyl and 1-Fluoronaphthalene
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).