Adjustable Height Base Stands, Labconco®
 :   Labconco
Adjustable Height Base Stands, Labconco®
 :  Labconco
Adjustable Height Base Stands, Labconco®
Adjustable Height Base Stands, Labconco®
For use with Basic™ 47, Basic 70, Fiberglass 30, Protector® XStream®, Protector Premier™, Protector ClassMate®, Protector PVC Perchloric Acid, Protector Stainless Steel Perchloric Acid, Logic+, Logic, Cell Logic, PuriCare Procedure Stations and Protector Radioisotope laboratory hoods
  • Telescoping Base Stands with fixed feet can be adjusted to eight height positions prior to installation
  • Telescoping legs are adjustable in 1" increments from 27.5" to 34.5"
  • Durable 1.75" tubular, epoxy-coated steel frame
Base stands are constructed of durable 4.5cm (13/4") tubular, glacier white epoxy-coated steel and support loads up to 453.6kg (1000lbs.). ADA-compliant. Stands with telescoping legs are adjustable from 69.9–85.1cm (271/2–331/2") high. Hydraulic lift base stands adjust from 64.8–85.1cm (251/2–331/2") high and include four powder-coated, die-cast feet with levelers. Manual hydraulic lift models include front-accessible hand crank. Electric hydraulic lift models include a front-accessible up/down switch.
 : Electrical components of hydraulic lift stands are UL listed.
 : Hoods can be placed on a base stand or base cabinet. Several base cabinet models are available: standard storage (see 46610-404 series), acid storage (see 46610-412 series), and solvent storage (see 46610-440 series).
Product Family Options
  • Désignation
    Electrical
    For Use With
    Profondeur
    Hauteur
    Width
    Outside Depth
    Outside Height
    REDISHIP
    Shipping Weight
  • 4' Telescoping Base Stand with Fixed Feet
    Logic+, Logic, Cell Logic Biosafety Cabinets and PuriCare Procedure Stations
    29"
    34.5"
    4 ft.
    Yes
    53 kg (117 lbs.)
     : 37304003726
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    3' Models
    29"
    27½ - 33½"
    72 lbs.
    Each
    Each - $1,790.00
    $1,790.00
    Each
       $1,790.00
     : 3746701
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    5' Models
    29"
    27½ - 33½"
    85 lbs.
    Each
    Each - $1,990.00
    $1,990.00
    Each
       $1,990.00
     : 3746703
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    6' Model
    29"
    27½ - 33½"
    92 lbs.
    Each
    Each - $2,100.00
    $2,100.00
    Each
       $2,100.00
     : 3746704
  • Adjustable Base Stand, Manual Hydraulic Lift
    3' Models
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $5,430.00
    $5,430.00
    Each
       $5,430.00
     : 3780400
  • Adjustable Base Stand, Manual Hydraulic Lift
    4' Models
    29"
    25½ - 33½"
    180 lbs.
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
     : 3780401
  • Adjustable Base Stand, Manual Hydraulic Lift
    6' Model
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $5,600.00
    $5,600.00
    Each
       $5,600.00
     : 3780402
  • Adjustable Base Stand, Electric Hydraulic Lift
    115 V, 60 Hz
    3' Models
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $6,840.00
    $6,840.00
    Each
       $6,840.00
     : 3780300
  • Adjustable Base Stand, Electric Hydraulic Lift
    115 V, 60 Hz
    4' Models
    29"
    25½ - 33½"
    180 lbs.
    Each
    Each - $6,930.00
    $6,930.00
    Each
       $6,930.00
     : 3780301
  • Adjustable Base Stand, Electric Hydraulic Lift
    115 V, 60 Hz
    6' Model
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $7,010.00
    $7,010.00
    Each
       $7,010.00
     : 3780302
  • Adjustable Base Stand, Electric Hydraulic Lift
    230 V, 50 Hz
    3' Models
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $6,840.00
    $6,840.00
    Each
       $6,840.00
     : 3780303
  • Adjustable Base Stand, Electric Hydraulic Lift
    230 V, 50 Hz
    4' Models
    29"
    25½ - 33½"
    180 lbs.
    Each
    Each - $6,930.00
    $6,930.00
    Each
       $6,930.00
     : 3780304
  • Adjustable Base Stand, Electric Hydraulic Lift
    230 V, 50 Hz
    6' Model
    29"
    25½ - 33½"
    150 lbs.
    Each
    Each - $7,010.00
    $7,010.00
    Each
       $7,010.00
     : 3780305
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    4' Models
    29"
    27½ - 33½"
    80 lbs.
    Each
    Each - $1,890.00
    $1,890.00
    Each
       $1,890.00
     : 3746702
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    3' Models
    29"
    27½ - 33½"
    72 lbs.
    Each
    Each - $1,790.00
    $1,790.00
    Each
       $1,790.00
     : 3746701
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    4' Models
    29"
    27½ - 33½"
    80 lbs.
    Each
    Each - $1,890.00
    $1,890.00
    Each
       $1,890.00
     : 3746702
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    6' Model
    29"
    27½ - 33½"
    92 lbs.
    Each
    Each - $2,100.00
    $2,100.00
    Each
       $2,100.00
     : 3746704
Accessories
  •  
    Description
  • 97006-416
    Accessory Shelf, 4'
    Each - $387.00
  •  
    Description
    For Use With
    Interior Dimensions
  • 97010-022
    Seismic Support Bracket Kit, for Telescoping Base Stands with Fixed Feet
    Each - $567.00
  •  
    Description
  • 97006-418
    Accessory Shelf, 5'
    Each - $483.00
  •  
    Description
  • 97006-420
    Accessory Shelf, 6'
    Each - $588.00
  •  
    Description
  • 97006-414
    Accessory Shelf, 3'
    Each - $285.00