VWR® Bath Fluids
VWR® Bath Fluids
VWR® Bath Fluids
VWR® Bath Fluids
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
  • Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
 : Bottles are packaged in a carton for shipment.
Product Family Options
  • Désignation
    Taille
    For Use With
    Temperature Range
    Capacité de flacon
  • PolyScience polytherm S250
    3.8 l (1 gal.)
    VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
    150...250 °C (302...482 °F)
    (25°C) 500 cSt
    Each (1US Gal)
    Each (1US Gal) - $3,876.06
    $3,876.06
    Each (1US Gal)
       $3,876.06
  • PolyScience polytherm S150
    3.8 l (1 gal.)
    VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
    50...150 °C (122...302 °F)
    (25°C) 50 cSt
    Each (1US Gal)
    Each (1US Gal) - $2,893.65
    $2,893.65
    Each (1US Gal)
       $2,893.65
  • PolyScience polycool HC -50, Freezing Point –56 °C
    3.8 l (1 gal.)
    VWR Refrigerated Circulating Baths
    –50...+100 °C (–58...+212 °F)
    (0°C) 3.6 cSt
    (–40°C) 15.1 cSt
    Each (1US Gal)
    Each (1US Gal) - $424.81
    $424.81
    Each (1US Gal)
       $424.81