Nalgene® Centrifuge Tubes, Polycarbonate, Conical-Bottom, Graduated, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
These tubes are ideal for whole cell separations.
- Maximum RCF: 6000 ×g
- Autoclavable
They are transparent, and have molded graduations. For proper performance, fill to 80% of total capacity.
Ordering information: Order closures separately.
Frequently Bought Together