NALGENE® Centrifuge Tube Closures, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Friction-fit polypropylene closures designed for NALGENE® round or conical centrifuge tubes
Pull tab for closure removal. Autoclavable. May not provide liquid-tight seal at high speeds. Closure 21010-965 is for use with round, nonlipped 50mL tubes.
Frequently Bought Together