VertiKlean® MAX™ Mopping Systems, Contec®
VertiKlean® MAX™ Mopping Systems, Contec®
Supplier:   Contec
VertiKlean® MAX™ Mopping Systems, Contec®
Supplier:  Contec
Certificates
VertiKlean® MAX™ Mopping Systems, Contec®
VertiKlean® MAX™ Mopping Systems, Contec®
13 Options Available Below
Product Details & Documents
VertiKlean® MAX™ Mop was designed for applying disinfectants to both horizontal and vertical surfaces, and is ideal for general cleaning of ceilings, walls, and floors in the most critical environments. This durable, lightweight mop features a single layer hydrophilic polyurethane foam laminated to polyester microfiber fabric, backed by a semi-rigid thermoformed plate, which easily snaps onto a standard VertiKlean® mop frame.
  • Hydrophilic polyurethane foam and knitted polyester mop head retains liquids
  • When the mop is applied to a surface, the liquid is evenly released without dripping
  • Laser cutting seals the edges of the foam and polyester, reducing the chance of fiber and particle contamination
  • VertiKlean® MAX™ Mop enables the uniform application of disinfectants and cleaning solutions
  • Excellent chemical and abrasion resistance
  • Lasts longer than traditional sponge mops
  • Compatible with a wide range of solutions
  • All three sizes available with a sealed edge
  • Available gamma irradiated. Validated sterile to a Sterility Assurance Level of 10⁻⁶
VertiKlean® MAX™ Mop has a slim profile for cleaning in confined spaces. Ribbed microfiber material smoothly glides over floors and textured surfaces and provides superior contamination removal. The pivoting mop frame makes it easy to clean around door and window frames as well as light fixtures, and heavy equipment. Available in three lengths to meet any cleaning needs, the smallest VertiKlean® MAX™ Mop is appropriate for cleaning tasks in confined spaces, such as cleaning and disinfecting fume hoods, isolators and mini environments.
The VertiKlean® MAX™ Mop Medium and Large are ideal for cleaning and disinfecting broad surfaces in cleanrooms and controlled environments. All three sizes are available with a sealed edge. For superior particle pick up and contamination removal, as well as maximum sorbency, choose VertiKlean® MAX™ for cleaning and disinfecting.
Product Family Options
Regular system
Product Information
  • Désignation
    Taille
    Sterility
    Packed
    Availability
    Price
  • Regular VertiKlean® MAX™ Ribbed Microfiber Mop Head
    20.3×10.2×2.5 cm (8×4×1")
    Non sterile
    4/Bag, 12 Bags/Case
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $858.28
    $858.28
    New Customer Promotional Price
    Case of 48
       $858.28
    Supplier Number: VKMX0125
  • Regular VertiKlean® MAX™ Sterile Ribbed Microfiber Mop Head
    20.3×10.2×2.5 cm (8×4×1")
    Sterile
    4/Bag, 12 Bags/Case
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $1,917.80
    $1,917.80
    New Customer Promotional Price
    Case of 48
       $1,917.80
    Supplier Number: VKMX0175
  • Regular VertiKlean® MAX™ Sealed Edge Mop Head
    24×12.7×1 cm (9.5×5×0.375")
    Non sterile
    4/Bag, 10 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $712.08
    $712.08
    New Customer Promotional Price
    Case of 40
       $712.08
    Supplier Number: VKSE0100
  • Regular VertiKlean® MAX™ Sterile Sealed Edge Mop Head
    24×12.7×1 cm (9.5×5×0.375")
    Sterile
    1/Bag, 40 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $885.80
    $885.80
    New Customer Promotional Price
    Case of 40
       $885.80
    Supplier Number: VKSE0150
Large system
Product Information
  • Désignation
    Taille
    Sterility
    Packed
    Availability
    Price
  • Large VertiKlean® MAX™ Ribbed microfiber mop head
    38×10.2×2.5 cm (15×4×1")
    Non sterile
    4/bag, 6 bags/case
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $946.00
    $946.00
    New Customer Promotional Price
    Case of 24
       $946.00
    Supplier Number: VKMX0325
  • Large VertiKlean® MAX™ Sterile Ribbed Microfiber Mop Head
    38×10.2×2.5 cm (15×4×1")
    Sterile
    1/Bag, 24 Bags/Case
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,728.60
    $1,728.60
    New Customer Promotional Price
    Case of 24
       $1,728.60
    Supplier Number: VKMX0375
  • Large VertiKlean® MAX™ Sealed Edge Mop Head
    46×13×1 cm (18×5×0.375")
    Non sterile
    4/Bag, 5 Bags/Case
    Discontinued
    Supplier Number: -
  • Large VertiKlean® MAX™ Sealed Edge Mop Head
    45.7×12.7×1 cm (18×5×0.375")
    Non sterile
    4/Bag, 5 Bags/Case
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $903.00
    $903.00
    New Customer Promotional Price
    Case of 20
       $903.00
    Supplier Number: VKSE0300
  • Large VertiKlean® MAX™ Sterile Sealed Edge Mop Head
    45.7×12.7×1 cm (18×5×0.375")
    Sterile
    1/Bag, 20 Bags/Case
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $1,358.80
    $1,358.80
    New Customer Promotional Price
    Case of 20
       $1,358.80
    Supplier Number: VKSE0350
Mop head frames
Product Information
  • Désignation
    Taille
    Sterility
    Packed
    Availability
    Price
  • QuickConnect™ Mop Frame, Stainless Steel
    19×4.4×10.8 cm (7.5×1.75×4.25")
    Discontinued
    Supplier Number: -
  • QuickConnect™ Mop Frame for Regular Systems
    15.2 cm (6")
    Log in for availability
    Log in for availability
    Each
    Each - $310.46
    $310.46
    New Customer Promotional Price
    Each
       $310.46
    Supplier Number: 2691A
  • QuickConnect™ Mop Frame, Stainless Steel
    17.8 cm (7")
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $227.90
    $227.90
    New Customer Promotional Price
    Log in for availability
    Each
    Each - $227.90
    $227.90
    New Customer Promotional Price
    Case of 1
       $227.90
    Each
       $227.90
    Supplier Number: 2651LF
  • QuickConnect™ Mop Frame for Regular Systems, Stainless Steel
    19 cm (7.5")
    Discontinued
    Supplier Number: -
  • QuickConnect™ Mop Frame for Medium & Large Systems, Stainless Steel
    29.2 cm (11.5")
    Log in for availability
    Log in for availability
    Each
    Each - $321.64
    $321.64
    New Customer Promotional Price
    Each
       $321.64
    Supplier Number: 2652SS
  • QuickConnect™ Flat Mop Frame for Medium & Large Systems, Stainless Steel
    29.2 cm (11.5")
    Log in for availability
    Log in for availability
    Each
    Each - $394.74
    $394.74
    New Customer Promotional Price
    Each
       $394.74
    Supplier Number: 2652LF
  • QuickConnect™ Mop Frame for Large Systems, Stainless Steel
    35.6 cm (14")
    Log in for availability
    Log in for availability
    Each
    Each - $330.24
    $330.24
    New Customer Promotional Price
    Each
       $330.24
    Supplier Number: 2653SS
  • QuickConnect™ Mop Frame for Large Systems
    35.6 cm (14")
    Discontinued
    Supplier Number: -
Medium system
Product Information
  • Catalog #
    Packed
    Taille
    Désignation
    Availability
    Price
  • 1/Bag, 28 Bags/Case
    33×10.2×2.5 cm (13×4×1")
    Medium VertiKlean® MAX™ Sterile ribbed microfiber mop head
    Log in for availability
    Log in for availability
    Case of 28
    Case of 28 - $1,634.00
    $1,634.00
    Case of 28
       $1,634.00
Product Information
  • Catalog #
    Packed
    Taille
    Désignation
    Availability
    Price
  • 4/Bag, 4 Bags/Case
    38×12.7×1 cm (15×5×0.375")
    Medium VertiKlean® MAX™ Sealed edge mop head
    Log in for availability
    Log in for availability
    Case of 16
    Case of 16 - $418.82
    $418.82
    Case of 16
       $418.82
Product Information
  • Catalog #
    Packed
    Taille
    Désignation
    Availability
    Price
  • 1/Bag, 24 Bags/Case
    38×12.7×1 cm (15×5×0.375")
    Medium VertiKlean® MAX™ Sterile sealed edge mop head
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $916.68
    $916.68
    Case of 24
       $916.68
Product Information
  • Catalog #
    Packed
    Taille
    Désignation
    Availability
    Price
  • 4/Bag, 7 Bags/Case
    33×10.2×2.5 cm (13×4×1")
    Medium VertiKlean® MAX™ Ribbed microfiber mop head
    Log in for availability
    Log in for availability
    Case of 28
    Case of 28 - $894.40
    $894.40
    Case of 28
       $894.40
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-717
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $761.10
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-715
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $265.74
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-716
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $502.24
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-714
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $235.64