EasyCurve™ Mop System, Contec® | Avantor
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
Supplier:  CONTEC, INC. MX
Certificates
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
5 Options Available Below
Product Details & Documents
The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.
  • All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal
The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability. This makes the EasyCurve™ an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve™ Mop are autoclavable. Available in both nonsterile and validated sterile versions.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • EasyCurve™ Ribbed Microfiber Mop Head
    36×11×2 cm (14×1.25×0.75")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $1,300.00
    $1,300.00
    Case of 48
       $1,300.00
    Supplier Number: EZMP0300
  • EasyCurve™ Sterile Ribbed Microfiber Mop Head
    36×11×2 cm (14×1.25×0.75")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $1,370.00
    $1,370.00
    Case of 48
       $1,370.00
    Supplier Number: EZMP0350
  • EasyCurve™ Sealed Edge Mop Head
    38×13×1 cm (15×5×0.375")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $1,049.20
    $1,049.20
    Prix promotionnel pour les nouveaux clients
    Case of 40
       $1,049.20
    Supplier Number: EZSE0100
  • EasyCurve™ Sterile Sealed Edge Mop Head
    38×13×1 cm (15×5×0.375")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $868.60
    $868.60
    Prix promotionnel pour les nouveaux clients
    Case of 24
       $868.60
    Supplier Number: EZSE0150
  • EasyCurve™ Stainless Steel Mop Frame
    36×11 cm (14×4.25")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $285.52
    $285.52
    Prix promotionnel pour les nouveaux clients
    Each
       $285.52
    Supplier Number: 2724
Accessories
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-715
    127 - 234 cm (50 - 92")
    Log in pour la disponibilité
    Each - $265.74
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-716
    41 - 76 cm (16 - 30")
    Log in pour la disponibilité
    Each - $502.24
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-714
    41 - 76 cm (16 - 30")
    Log in pour la disponibilité
    Each - $235.64
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stainless Steel Connecting Kit for Multi-Bucket System
    Stainless Steel Connecting Kit for Multi-Bucket System
    89073-986
    Log in pour la disponibilité
    Each - $127.28
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-717
    127 - 234 cm (50 - 92")
    Log in pour la disponibilité
    Each - $761.10