VWR® Angled Foam Wall Mop | Avantor
VWR® Angled Foam Wall Mop
Mop Head, 35.6 cm (14")
Catalog # 89012-766
Supplier:   VWR International
VWR® Angled Foam Wall Mop
Catalog # 89012-766
Supplier:  VWR International
VWR® Angled Foam Wall Mop
VWR® Angled Foam Wall Mop
Mop Head, 35.6 cm (14")
Price
$1,160.77
Log in pour la disponibilité
Case of 28
Product Details & Documents
Wedge-shaped mop head is made of knitted polyester fabric laminated to clean ester foam for excellent abrasion resistance.
  • Fully autoclavable
  • Stainless steel swivel-head frame
Ideal for cleaning ceilings and walls, and for applying disinfectants and cleaning solvents in controlled environments
Swivel-head frame is durable stainless steel. Features a capped end for maximum cleanliness and a quick-connect button for easy attachment to interchangeable handles.
Telescoping handles are available in two materials, lightweight anodized aluminum or durable stainless steel, and two lengths.
Large size allows efficient coverage of large areas, while wide leading edge and angled sides allow easy access to tight corners. Compatible with a wide range of chemicals. Autoclavable at 121°C (250°F) for 30 minutes. Suitable for ISO Class 5 (FED-STD-209D Class 100/M3.5) conditions. Fits a 30.5 cm (12") frame (not included).
Ordering information: Mop head, mop frame, and handle must all be purchased separately.
Documents
Spécifications
  • Désignation:
    Mop Head, 35.6 cm (14")
Accessories
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-717
    127 - 234 cm (50 - 92")
    Log in pour la disponibilité
    Each - $761.10
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-716
    41 - 76 cm (16 - 30")
    Log in pour la disponibilité
    Each - $502.24
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-715
    127 - 234 cm (50 - 92")
    Log in pour la disponibilité
    Each - $265.74
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-714
    41 - 76 cm (16 - 30")
    Log in pour la disponibilité
    Each - $235.64