VertiKlean® Wall Washing Systems
VertiKlean® Wall Washing Systems
Supplier:   Contec
VertiKlean® Wall Washing Systems
Supplier:  Contec
Certificates
VertiKlean® Wall Washing Systems
VertiKlean® Wall Washing Systems
13 Options Available Below
Product Details & Documents
Lightweight, polyester disposable mops are excellent for applying disinfectants and for wet cleaning of vertical surfaces, ceilings, and walls in controlled environments. Ideal for reaching awkward corners and ceilings with a complete range of motion.
  • Hydrophilic polyester fabric laminated to clean ester foam, laser cut sealed edge
  • Compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
  • Available in three sizes (8, 13 and 15")
For aseptic processing or sterile core areas, the sterile mop heads provide truly validated sterility to a Sterility Assurance Level of 10-6 in accordance with standards and recommended practices of the AAMI guidelines.
Mop heads are constructed with a polyester clean room knit fabric laminated to clean ester foam. The absorbent foam carries solvents and disinfectants to the surface; the polyester fabric offers excellent abrasion resistance and helps distribute the liquid uniformly on the surface being cleaned. Mop heads are laser cut to seal the edges, further limiting potential contamination in the mopping process. Mop heads with a textured surface are ideal for applications requiring additional scrubbing performance. Mop heads contain no cellulose and are compatible with a wide range of chemicals, detergents, disinfectants, and cleaners. Intended for one-time use. Mop heads are available nonsterile and sterile.
Interchangeable hardware system consists of a disposable polyester/foam mop head attached to a stainless steel swivel mop frame with a capped end. A 'quick connect' feature allows for easy attachment to compatible extension handles with a 360° range of cleaning motions.
The VertiKlean® Sox removable covers are designed to protect the disposable mop heads during vertical wet cleaning. They improve the durability of the mop while cleaning highly abrasive surfaces. Compatible with all three VertiKlean® mop heads, the stainless steel bucket systems include trolley, casters, buckets, and wringer. All units in the VertiKlean® system are compatible with gamma, ETO, and autoclave sterilization.
Ordering information: Validation reports and presaturated and sterile mop head covers are available upon special request. Paddle-style 41.3 cm (16¹/₄") long VertiKlean® mop heads are also available; contact your Avantor sales representative for more information. Mop heads are available in three sizes, 21.6 cm (8¹/₂"), 32.4 cm (12³/₄"), and 38.7 cm (15¹/₄"); for use with their corresponding 19 cm (7 or 7¹/₂"), 29 cm (11¹/₂ or 12"), and 36 cm (14") frames, respectively. Lightweight anodized aluminum and stainless steel telescoping handles are available in two lengths: 40 cm (16") extending to 76.2 cm (30"), and 127 cm (50") extending to 233.7 cm (92").
Product Family Options
Regular System
Product Information
  • Désignation
    Matière
    Sterility
    Availability
    Price
  • Standard mop head, 21.6 cm (8½") long, textured
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $699.18
    $699.18
    New Customer Promotional Price
    Case of 48
       $699.18
    Supplier Number: TVKMOP
  • VertiKlean® Sox removable cover, for 21.6 cm (8½") mop head
    Polyester
    Non sterile
    Discontinued
    Supplier Number: -
  • Standard mop head, 21.6 cm (8½") long
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $220.16
    $220.16
    New Customer Promotional Price
    Log in for availability
    Case of 48
    Case of 48 - $880.64
    $880.64
    New Customer Promotional Price
    Pack of 12
       $220.16
    Case of 48
       $880.64
    Supplier Number: VK-MOP
  • Standard mop head, 17.8 cm (7") long, sterile
    Polyester
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $1,823.20
    $1,823.20
    New Customer Promotional Price
    Log in for availability
    Pallet of 864
    Pallet of 864 - $32,817.60
    $32,817.60
    New Customer Promotional Price
    Case of 48
       $1,823.20
    Pallet of 864
       $32,817.60
    Supplier Number: VK-MOPIR
Medium System
Product Information
  • Désignation
    Matière
    Sterility
    Availability
    Price
  • VertiKlean® Sox Quiltec® removable cover, for 32.4 cm (12¾") mop head
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $1,135.20
    $1,135.20
    New Customer Promotional Price
    Case of 12
       $1,135.20
    Supplier Number: MVK-QT-SOX
  • Medium VertiKlean® Mop Head, 12.75×4.25×1", textured
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $963.20
    $963.20
    New Customer Promotional Price
    Case of 32
       $963.20
    Supplier Number: TMVKMOP
  • Standard mop head, 32.4 cm (12¾") long
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $92.02
    $92.02
    New Customer Promotional Price
    Log in for availability
    Case of 32
    Case of 32 - $736.16
    $736.16
    New Customer Promotional Price
    Pack of 4
       $92.02
    Case of 32
       $736.16
    Supplier Number: MVK-MOP
  • Standard mop head, 32.4 cm (12¾") long, sterile
    Polyester
    Sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $1,651.20
    $1,651.20
    New Customer Promotional Price
    Case of 32
       $1,651.20
    Supplier Number: MVK-MOPIR
  • Standard mop head, 32.4 cm (12¾") long, textured, sterile
    Polyester
    Sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $1,419.00
    $1,419.00
    New Customer Promotional Price
    Case of 32
       $1,419.00
    Supplier Number: TMVKMOPIR
Large System
Product Information
  • Désignation
    Matière
    Sterility
    Availability
    Price
  • VertiKlean® Sox removable cover, for 38.7 cm (15¹/₄") mop head
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $345.72
    $345.72
    New Customer Promotional Price
    Case of 32
       $345.72
    Supplier Number: LVK-SOX
  • Standard mop head, 38.7 cm (15¹/₄") long, textured
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $954.60
    $954.60
    New Customer Promotional Price
    Case of 32
       $954.60
    Supplier Number: TLVKMOP
  • Standard mop head, 38.7 cm (15¹/₄") long
    Polyester
    Non sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $106.64
    $106.64
    New Customer Promotional Price
    Log in for availability
    Case of 32
    Case of 32 - $853.12
    $853.12
    New Customer Promotional Price
    Pack of 4
       $106.64
    Case of 32
       $853.12
    Supplier Number: LVK-MOP
  • Standard mop head, 38.7 cm (15¹/₄") long, textured, sterile
    Polyester
    Sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $1,608.20
    $1,608.20
    New Customer Promotional Price
    Case of 32
       $1,608.20
    Supplier Number: TLVKMOPIR
  • Standard mop head, 38.7 cm (15¹/₄") long, sterile
    Polyester
    Sterile
    Log in for availability
    Log in for availability
    Case of 32
    Case of 32 - $1,513.60
    $1,513.60
    New Customer Promotional Price
    Case of 32
       $1,513.60
    Supplier Number: LVK-MOPIR
Mop Head Frames
Product Information
  • Catalog #
    Taille
    Désignation
    Availability
    Price
  • 15.2 cm (6")
    QuickConnect™ Mop Frame for Regular Systems
    Log in for availability
    Log in for availability
    Each
    Each - $310.46
    $310.46
    Each
       $310.46
QuickConnect™ Handles
Product Information
  • Catalog #
    Désignation
    Longueur
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    127 - 234 cm (50 - 92")
    Log in for availability
    Log in for availability
    Each
    Each - $265.74
    $265.74
    Each
       $265.74
Product Information
  • Catalog #
    Désignation
    Longueur
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    41 - 76 cm (16 - 30")
    Log in for availability
    Log in for availability
    Each
    Each - $502.24
    $502.24
    Each
       $502.24
Product Information
  • Catalog #
    Désignation
    Longueur
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    127 - 234 cm (50 - 92")
    Log in for availability
    Log in for availability
    Each
    Each - $761.10
    $761.10
    Each
       $761.10
Product Information
  • Catalog #
    Désignation
    Longueur
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    41 - 76 cm (16 - 30")
    Log in for availability
    Log in for availability
    Each
    Each - $235.64
    $235.64
    Each
       $235.64
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Anodized Aluminum Handle, 2 Piece, 218 cm (86") Handle, 30 cm (12") Frame
    Anodized Aluminum Handle, 2 Piece, 218 cm (86") Handle, 30 cm (12") Frame
    33502-010
    Log in for availability
    Each - $280.36