Continuous Extractor, Liquid-Liquid, Chemglass
Continuous Extractor, Liquid-Liquid, Chemglass
Extractor Only
Catalog # 80068-146
Supplier:   Chemglass
Supplier #:  CG-1360-01
Continuous Extractor, Liquid-Liquid, Chemglass
Catalog # 80068-146
Supplier:  Chemglass
Continuous Extractor, Liquid-Liquid, Chemglass
Continuous Extractor, Liquid-Liquid, Chemglass
Extractor Only
Price
$1,090.00
Log in for availability
Each
Product Details & Documents
Designed for EPA priority pollutant samples where the extracting solvent is heavier than water
For use with samples of about 1L plus approximately 200mL of extracting solvent. Extractor has a [ST] 45/50 top outer joint and a [ST] 24/40 lower inner joint.
Ordering information: Accessories sold separately.
Specifications
  • Désignation:
    Extractor Only
Accessories
Product Information
  • Catalog #
    Capacity
    ST Joint No.
    Availability
    Price
  • Flask
    Flask
    80068-786
    1 L
    24/40
    Log in for availability
    Each - $123.00
Product Information
  • Catalog #
    Availability
    Price
  • Condenser Only, Medium
    Condenser Only, Medium
    80068-160
    Log in for availability
    Each - $358.00