Starsys® Mobile Workstations, Metro™
Workstation components are constructed of corrosion-proof polymer for a contamination-free product.
- The countertops are 1" (25 mm) thick black epoxy resin
- Overhead units include 27" (686 mm) cabinets with locking clear doors and two poly shelves
Microban® Antimicrobial product protection inhibits the growth of stain and odor-causing bacteria on Starsys, keeping the surface areas cleaner between cleanings. All preconfigured units come standard with taupe door/drawer pulls and polymer 5" (127 mm) Ø casters (2-total lock, 2-swivel).
Frequently Bought Together