Variable-Speed Electric Stirrers, Arrow | Avantor
Variable-Speed Electric Stirrers, Arrow
Supplier:   Arrow Engineering
Variable-Speed Electric Stirrers, Arrow
Supplier:  Arrow Engineering
Certificates
Variable-Speed Electric Stirrers, Arrow
4 Options Available Below
Product Details & Documents
These variable-speed, electronically controlled stirrers are built to handle viscosities ranging from water-like solutions to highly viscous fluids
Four models are available to handle a wide range of applications: Model 6000 is a low-cost, light-duty stirrer, ideal for stirring light to medium-light viscosity fluids. Model 1750 is a multi-purpose stirrer, designed to handle more viscous materials. Model 850 provides the speed and power to handle highly viscous materials. Its high-torque gear head maintains constant speed regardless of viscosity changes as mixing progresses. Slim design is ideal for use in three-neck flasks. Model 350 is a high-torque, low-rpm stirrer, specifically designed to maintain constant low speed. Maximum volume that stirrers can handle is dependent on liquid viscosity, shaft length, and stirring speed.
Totally enclosed, non-ventilated motor provides maximum constant torque throughout entire speed range. Eye-level, heavy cast-aluminum control box integrated with motor contains control knob for electronically regulating stirrer speed and on/off switch. Stainless steel shaft and propeller resist most acids and chemicals for easy cleaning.
Ordering information: Complete with 25.4L x 1.4dia. cm (10 x 9/16") aluminum support rod and 1.8m (6') three-wire cord with grounded plug. Shaft measures 30.5L x 1dia. cm (12 x 3/8") and has a 6.4cm (21/2") diameter propeller.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 59025-124
  • 59025-126
  • 59025-128
  • 59025-130
  • Max. stirring capacity H₂O:
  • 20 L
  • 100 L
  • 100 L
  • 50 L
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Electrical
    Speed Range
    Horsepower
    Shipping Weight
    Torque
    Availability
    Price
  • Model 6000, Light-Duty; Gear Ratio: None
    115 V, 60 Hz/230 V, 50 Hz
    0 - 6000 rpm
    ⅒ hp W
    3.6 kg (7.9 lbs.)
    1.1 inc.Ibs..
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,504.42
    $2,504.42
    Each
       $2,504.42
    Supplier Number: ARROW6000
    Spécifications:
    CSA Certified
    More
    Specifications
    Cat. No.
    59025-124
    Max. stirring capacity H₂O
    20 L
    CSA Certified
  • Model 1750, General Purpose, Gear Ratio: None
    115 V, 60 Hz/230 V, 50 Hz
    0 - 2000 rpm
    ¹⁄₁₅ hp W
    3.6 kg (7.9 lbs.)
    2.4 inc.Ibs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,571.86
    $2,571.86
    Each
       $2,571.86
    Supplier Number: ARROW1750
    Spécifications:
    CSA Certified
    More
    Specifications
    Cat. No.
    59025-126
    Max. stirring capacity H₂O
    100 L
    CSA Certified
  • Model 850, Heavy-Duty, Gear Ratio: 7 to 1
    115 V, 60 Hz/230 V, 50 Hz
    0 - 1000 rpm
    ⅒ hp W
    4.1 kg (9 lbs.)
    7.3 inc.Ibs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,197.25
    $3,197.25
    Each
       $3,197.25
    Supplier Number: ARROW850
    Spécifications:
    CSA Certified
    More
    Specifications
    Cat. No.
    59025-128
    Max. stirring capacity H₂O
    100 L
    CSA Certified
  • Model 350, Heavy-Duty, Gear Ratio: 7 to 1
    115 V, 60 Hz/230 V, 50 Hz
    0 - 350 rpm
    ¹⁄₁₅ hp W
    4.1 kg (9 lbs.)
    17.1 inc.Ibs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,305.60
    $3,305.60
    Each
       $3,305.60
    Supplier Number: ARROW350-115
    Spécifications:
    CSA Certified
    More
    Specifications
    Cat. No.
    59025-130
    Max. stirring capacity H₂O
    50 L
    CSA Certified