Protector® Solvent Storage Cabinets, Labconco®
About this item
Safely store and vent solvents and other highly flammable liquids.
- Dual-wall white epoxy-coated steel construction with 3.8 cm (³/₂") air gap for complete protection
Equipped with three-point locking handle, rear vent outlets with flame arresters and closure plugs, dished epoxy-coated steel shelf, and leveling legs. Include one 20.3 cm (8") filler panel to increase cabinet's overall depth from 55.9 cm (22") to 76.2 cm (30"), for installation flush to wall.
Certifications: Meets OSHA and NFPA standards for storage cabinets. Self-closing cabinets meet UFC97.
Ordering information: A work surface (sold separately) is required for use with these cabinets, when used to support hoods without integral bottoms. The solvent vent kit includes a flexible metal hose 3.8 cm (³/₂") in diameter, two hose clamps, and 5.1×3.8 cm (2׳/₂") NPT slip-fit outlet adapter.
Delivery information: Items designated as REDISHIP can be shipped within 48 hours of receiving your order.
15 Options Available Below
Product Details & Documents
Safely store and vent solvents and other highly flammable liquids.
- Dual-wall white epoxy-coated steel construction with 3.8 cm (³/₂") air gap for complete protection
Equipped with three-point locking handle, rear vent outlets with flame arresters and closure plugs, dished epoxy-coated steel shelf, and leveling legs. Include one 20.3 cm (8") filler panel to increase cabinet's overall depth from 55.9 cm (22") to 76.2 cm (30"), for installation flush to wall.
Certifications: Meets OSHA and NFPA standards for storage cabinets. Self-closing cabinets meet UFC97.
Ordering information: A work surface (sold separately) is required for use with these cabinets, when used to support hoods without integral bottoms. The solvent vent kit includes a flexible metal hose 3.8 cm (³/₂") in diameter, two hose clamps, and 5.1×3.8 cm (2׳/₂") NPT slip-fit outlet adapter.
Delivery information: Items designated as REDISHIP can be shipped within 48 hours of receiving your order.
Product Family Options
Product Information
- DésignationTypeLargeurProfondeurHauteurShipping WeightREDISHIPAvailabilityPrice
- Solvent storage cabinet, ADA height1 automatic self-closing door, left hinge2 ft.22"35½ - 36¾"160.0 lbsNoLog in for availabilityEach$5,810.00Supplier Number: 9906300
- Solvent storage cabinet, standard height1 automatic self-closing door, left hinge2 ft.22"35½ - 36¾"165.0 lbsNoLog in for availabilityEach$5,610.00Supplier Number: 9903400
- Solvent storage cabinet, ADA height1 automatic self-closing door, right hinge2 ft.22"35½ - 36¾"150.0 lbsNoLog in for availabilityEach$5,810.00Supplier Number: 9906200
- Solvent storage cabinet, standard height1 automatic self-closing door, right hinge2 ft.22"35½ - 36¾"165.0 lbsNoLog in for availabilityEach$5,610.00Supplier Number: 9903300
- Solvent storage cabinet, standard height2 automatic self-closing doors2½ ft.22"35½ - 36¾"215.0 lbsNoLog in for availabilityEach$6,490.00Supplier Number: 9903200
- Solvent storage cabinet, standard height2 automatic self-closing doors3 ft.22"35½ - 36¾"225.0 lbsNoLog in for availabilityEach$6,630.00Supplier Number: 9903100
- Solvent storage cabinet, standard height2 automatic self-closing doors4 ft.22"35½ - 36¾"270.0 lbs.NoLog in for availabilityEach$6,890.00Supplier Number: 9903000
- Solvent storage cabinet, ADA height1 manual door, left hinge2 ft.22"35½ - 36¾"160.0 lbsNoLog in for availabilityEach$4,970.00Supplier Number: 9906100
- Solvent storage cabinet, standard height1 manual door, left hinge2 ft.22"35½ - 36¾"155.0 lbsNoLog in for availabilityEach$4,810.00Supplier Number: 9902400
- Solvent storage cabinet, ADA height1 manual door, right hinge2 ft.22"35½ - 36¾"150.0 lbsNoLog in for availabilityEach$4,970.00Supplier Number: 9906000
- Solvent storage cabinet, standard height1 manual door, right hinge2 ft.22"35½ - 36¾"155.0 lbsNoLog in for availabilityEach$4,810.00Supplier Number: 9902300
- Solvent storage cabinet, standard height2 manual doors2½ ft.22"35½ - 36¾"195.0 lbsYesLog in for availabilityEach$4,920.00Supplier Number: 9902200
- Solvent storage cabinet, standard height2 manual doors3 ft.22"35½ - 36¾"205.0 lbsYesLog in for availabilityEach$5,080.00Supplier Number: 9902100
- Solvent storage cabinet, standard height2 manual doors4 ft.22"35½ - 36¾"250.0 lbsYesLog in for availabilityEach$5,320.00Supplier Number: 9902000
- Solvent storage cabinet, standard height1 manual door, right hinge2 ft.22"35½ - 36¾"155.0 lbsNoLog in for availabilityEach$4,810.00Supplier Number: 9902300
Accessories
Product Information
- Catalog #HeightShipping WeightAvailabilityPrice
- Solvent vent kit