Perforated Top Clean Tables, Metro™
Supplier: Metropolitan Wire
Certificates
About this item
These clean tables are made of type 304 stainless steel with HD super posts and super Erecta® construction throughout.
- The perforated top is manufactured from 14-gauge
- Support members have a satin finish
Type 304 stainless steel with 1.9 cm (³/₄") holes on 2.5 cm (1") centers. These units provide approximately 35% open area.
Perforated top tables are available in the overhead style with 188 cm (74") high rear posts. Use the cantilever shelves, grid panels and accessories to customize the work area.
All perforated top tables feature height adjustable legs 83.8 to 101.6 cm (33 to 40") in 2.5 cm (1") increments. Shipped knocked down, the units can be easily assembled in minutes.
Frequently Bought Together