Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
17L x 9W x 4H cm (611/16 x 31/2 x 19/16")
Catalog # 89009-084
Supplier: Atago
: 3464
Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
Catalog # 89009-084
Supplier: Atago
Jump to:
Product Details & Documents
Model UG-[alpha] is a compact urine specific gravity refractometer with digital display
Simple operation involves placing sample onto prism and pressing start. Measurements will stream across the LCD display. The minimum scale indication displays up to the fourth decimal. The UG-[alpha] refractometer is equipped with Automatic Temperature Compensation to provide accurate measurements, regardless of environmental conditions.
: Uses one 9V battery.
Spécifications
- Dimensions:17L x 9W x 4H cm (611/16 x 31/2 x 19/16")
- Measurement range:1.0000–1.0600 Urine S.G.
- Cat. No.:89009-084
- Measurement temperature range:10 to 35°C (50 to 95°F)
- Measurement accuracy:±0.0010 Urine S.G.
- Min. indication:0.0001 Urine S.G.
- Weight:300 g (10.6 oz.)