MiniSpin®/MiniSpin® plus Mini Centrifuge
Supplier:   Eppendorf
MiniSpin®/MiniSpin® plus Mini Centrifuge
Supplier:  Eppendorf
Certificates
MiniSpin®/MiniSpin® plus Mini Centrifuge
2 Options Available Below
Product Details & Documents
Eppendorf MiniSpin® and MiniSpin® plus centrifuges are powerful, entry-level, easy-to-use centrifuges that are small enough to allow individual workstations to be equipped with their own. Key parts are made of metal for maximum safety, and the MiniSpin® plus is fast enough for molecular biology separations.
  • Acceleration and deceleration time <13 sec
  • Metal rotor bowl
  • Separate short-spin key for fast and comfortable centrifugation
  • Optimal ventilation flow reduces heating and protects temperature-sensitive samples
  • Soft-touch lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
Special features for MiniSpin plus:
Automatic rpm/rcf conversion.
Extended time to 99 min or continous centrifugation.
 : Certificate of quality/conformity (ISO 9001, ISO 13485, ISO 14001).
 : Warranty: 2-Years Manufacturer's limited warranty. For a detailed warranty statement, please visit Eppendorf US website.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA47727-632
  • CA47727-636
  • Max. capacity:
  • 12×1.5/2.0 ml tubes (including rotor)
  • 12×1.5/2.0 ml (including rotor)
  • Max. speed:
  • 13400 rpm
  • 14500 rpm
  • Max. RCF:
  • 12100×g
  • 14100×g
  • Refrigerated:
  • No
  • No
  • Acceleration/braking time:
  • 13/12 s
  • 13/12 s
  • Control:
  • Keypad
  • Keypad
  • Electrical:
  • 120 V, 50/60 Hz
  • 120 V, 50/60 Hz
  • Dimensions:
  • 22.5×23.0×13.0 cm (8.86×9.06×5.12")
  • 22.5×23.0×13.0 cm (8.86×9.06×5.12")
  • Weight:
  • 4.1 kg
  • 4.1 kg
Spécifications
Product Family Options
MiniSpin®
Informations sur le produit
  • Désignation
    Rotors Included
    Availability
    Price
  • MiniSpin® centrifuge
    F-45-12-11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,810.00
    $2,810.00
    Each
       $2,810.00
    Supplier Number: 022620100
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47727-632
    Max. capacity
    12×1.5/2.0 ml tubes (including rotor)
    Max. speed
    13400 rpm
    Max. RCF
    12100×g
    Refrigerated
    No
    Acceleration/braking time
    13/12 s
    Control
    Keypad
    Electrical
    120 V, 50/60 Hz
    Dimensions
    22.5×23.0×13.0 cm (8.86×9.06×5.12")
    Weight
    4.1 kg
MiniSpin® plus
Informations sur le produit
  • Désignation
    Rotors Included
    Availability
    Price
  • MiniSpin® plus centrifuge
    F-45-12-11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,280.00
    $3,280.00
    Each
       $3,280.00
    Supplier Number: 022620207
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47727-636
    Max. capacity
    12×1.5/2.0 ml (including rotor)
    Max. speed
    14500 rpm
    Max. RCF
    14100×g
    Refrigerated
    No
    Acceleration/braking time
    13/12 s
    Control
    Keypad
    Electrical
    120 V, 50/60 Hz
    Dimensions
    22.5×23.0×13.0 cm (8.86×9.06×5.12")
    Weight
    4.1 kg
Rotors
Informations sur le produit
  • Catalog #
    Désignation
    Maximum RCF
    Maximum Capacity
    Maximum Speed
    Availability
    Price
  • F-55-16-5-PCR, includes rotor lid
    14100×g
    2×PCR strips
    14500 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $749.00
    $749.00
    Each
       $749.00
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Rotor lid for F-45-12-11, for MiniSpin/MiniSpin plus
    Rotor lid for F-45-12-11, for MiniSpin/MiniSpin plus
    75875-140
    Log in pour la disponibilité
    Each - $266.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • FA-24×2/F-32×0.2-PCR adapter, for 1×micro test tube (0.4 ml, max. Ø 6 mm), 6 pieces
    FA-24×2/F-32×0.2-PCR adapter, for 1×micro test tube (0.4 ml, max. Ø 6 mm), 6 pieces
    CA58922-858
    Log in pour la disponibilité
    Pack of 6 - $86.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • FA-24×2/F-32×0.2-PCR adapter for 0.5 ml micro-centrifuge tube
    FA-24×2/F-32×0.2-PCR adapter for 0.5 ml micro-centrifuge tube
    CA21516-180
    Log in pour la disponibilité
    Pack of 6 - $78.40
Informations sur le produit
  • Catalog #
    Availability
    Price
  • FA-24×2/F-32×0.2-PCR adapter, for 1×PCR tube (0.2 ml, max. Ø 6 mm), for all 1.5/2.0 ml rotors, 6 pieces, for all 1.5/2 ml rotors
    FA-24×2/F-32×0.2-PCR adapter, for 1×PCR tube (0.2 ml, max. Ø 6 mm), for all 1.5/2.0 ml rotors, 6 pieces, for all 1.5/2 ml rotors
    CA58922-860
    Log in pour la disponibilité
    Pack of 6 - $78.40