Lab Worktables, Mobile, Metro
Lab Worktables, Mobile, Metro
Supplier:   Metropolitan Wire
Lab Worktables, Mobile, Metro
Supplier:  Metropolitan Wire
Certificates
Lab Worktables, Mobile, Metro
Lab Worktables, Mobile, Metro
32 Options Available Below
Product Details & Documents
Stainless steel lab worktables provides corrosion-proof performance.
  • Tables feature smooth, easy-to-clean surfaces
  • Tops are made of Type 304 stainless steel or Trespa® phenolic resin
  • Phenolic resin tops are compression molded of organic fiber-reinforced phenolic core with an integrally cured thermoset resin surface
  • Phenolic resin tops are ideal for moderate to high chemical use and have excellent abrasion resistance and impact resistance
Tables are load rated at 244 kg/m2 (50lbs./ft.2) up to a maximum of 272 kg (600lbs.), assuming evenly distributed load and caster specifications that meet requirements. Tops with backsplash feature a 10.2cm (4") high backsplash.
Ordering information: Casters for mobile worktables must be ordered separately; for casters and other accessories, see 97000-762 series.
Product Family Options
Informations sur le produit
  • Désignation
    Worksurface Color/Finish
    Worksurface Material
    Base/Leg Material
    Hauteur
    W×D
    Shipping Weight
    Availability
    Price
  • Mobile worktables with solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    29¾×30"
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,489.02
    $3,489.02
    Each
       $3,489.02
    Supplier Number: LTSM30IS
  • Mobile worktables with solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    35¾×30"
    80 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,681.90
    $3,681.90
    Each
       $3,681.90
    Supplier Number: LTSM36IS
  • Mobile worktables with solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    47¾×30"
    103 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,937.02
    $3,937.02
    Each
       $3,937.02
    Supplier Number: LTSM48IS
  • Mobile worktables with solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    59¾×30"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,193.26
    $4,193.26
    Each
       $4,193.26
    Supplier Number: LTSM60IS
  • Mobile worktables with 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    29¾×30"
    58 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,489.02
    $3,489.02
    Each
       $3,489.02
    Supplier Number: LTSM30UIS
  • Mobile worktables with 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    35¾×30"
    70 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,681.90
    $3,681.90
    Each
       $3,681.90
    Supplier Number: LTSM36UIS
  • Mobile worktables with 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    47¾×30"
    91 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,937.02
    $3,937.02
    Each
       $3,937.02
    Supplier Number: LTSM48UIS
  • Mobile worktables with 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    59¾×30"
    119 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,193.26
    $4,193.26
    Each
       $4,193.26
    Supplier Number: LTSM60UIS
  • Mobile worktables with backsplash and solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    29¾×30"
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,489.02
    $3,489.02
    Each
       $3,489.02
    Supplier Number: LTSM30S
  • Mobile worktables with backsplash and solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    35¾×30"
    80 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,681.90
    $3,681.90
    Each
       $3,681.90
    Supplier Number: LTSM36S
  • Mobile worktables with backsplash and solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    47¾×30"
    103 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,937.02
    $3,937.02
    Each
       $3,937.02
    Supplier Number: LTSM48S
  • Mobile worktables with backsplash and solid HD shelf
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    59¾×30"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,193.26
    $4,193.26
    Each
       $4,193.26
    Supplier Number: LTSM60S
  • Mobile worktables with backsplash and 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    29¾×30"
    58 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,489.02
    $3,489.02
    Each
       $3,489.02
    Supplier Number: LTSM30US
  • Mobile worktables with backsplash and 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    35¾×30"
    70 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,681.90
    $3,681.90
    Each
       $3,681.90
    Supplier Number: LTSM36US
  • Mobile worktables with backsplash and 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    47¾×30"
    91 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,937.02
    $3,937.02
    Each
       $3,937.02
    Supplier Number: LTSM48US
  • Mobile worktables with backsplash and 3-sided frame
    Silver
    Stainless steel
    Stainless steel
    35³⁄₁₆"
    59¾×30"
    119 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,193.26
    $4,193.26
    Each
       $4,193.26
    Supplier Number: LTSM60US
  • Mobile worktables with solid HD shelf
    Black
    Phenolic resin
    Stainless steel
    35⅜"
    29¾×30"
    77 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,421.78
    $3,421.78
    Each
       $3,421.78
    Supplier Number: LTSM30PB
  • Mobile worktables with solid HD shelf
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    35¾×30"
    92 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,617.88
    $3,617.88
    Each
       $3,617.88
    Supplier Number: LTSM36PB
  • Mobile worktables with solid HD shelf
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    47¾×30"
    113 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,869.81
    $3,869.81
    Each
       $3,869.81
    Supplier Number: LTSM48PB
  • Mobile worktables with solid HD shelf
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    59¾×30"
    141 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,128.50
    $4,128.50
    Each
       $4,128.50
    Supplier Number: LTSM60PB
  • Mobile worktables with solid HD shelf
    Gray
    Phenolic resin
    Stainless steel
    35⅜"
    29¾×30"
    77 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,421.78
    $3,421.78
    Each
       $3,421.78
    Supplier Number: LTSM30PG
  • Mobile worktables with solid HD shelf
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    35¾×30"
    92 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,376.75
    $3,376.75
    Each
       $3,376.75
    Supplier Number: LTSM36PG
  • Mobile worktables with solid HD shelf
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    47¾×30"
    113 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,869.81
    $3,869.81
    Each
       $3,869.81
    Supplier Number: LTSM48PG
  • Mobile worktables with solid HD shelf
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    59¾×30"
    141 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,128.50
    $4,128.50
    Each
       $4,128.50
    Supplier Number: LTSM60PG
  • Mobile worktables with 3-sided frame
    Black
    Phenolic resin
    Stainless steel
    35⅜"
    29¾×30"
    70 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,421.78
    $3,421.78
    Each
       $3,421.78
    Supplier Number: LTSM30UPB
  • Mobile worktables with 3-sided frame
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    35¾×30"
    82 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,617.88
    $3,617.88
    Each
       $3,617.88
    Supplier Number: LTSM36UPB
  • Mobile worktables with 3-sided frame
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    47¾×30"
    101 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,869.81
    $3,869.81
    Each
       $3,869.81
    Supplier Number: LTSM48UPB
  • Mobile worktables with 3-sided frame
    Black
    Phenolic resin
    Stainless steel
    35⅞"
    59¾×30"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,128.50
    $4,128.50
    Each
       $4,128.50
    Supplier Number: LTSM60UPB
  • Mobile worktables with 3-sided frame
    Gray
    Phenolic resin
    Stainless steel
    35⅜"
    29¾×30"
    70 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,421.78
    $3,421.78
    Each
       $3,421.78
    Supplier Number: LTSM30UPG
  • Mobile worktables with 3-sided frame
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    35¾×30"
    82 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,378.70
    $3,378.70
    Each
       $3,378.70
    Supplier Number: LTSM36UPG
  • Mobile worktables with 3-sided frame
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    47¾×30"
    101 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,869.81
    $3,869.81
    Each
       $3,869.81
    Supplier Number: LTSM48UPG
  • Mobile worktables with 3-sided frame
    Gray
    Phenolic resin
    Stainless steel
    35⅞"
    59¾×30"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,128.50
    $4,128.50
    Each
       $4,128.50
    Supplier Number: LTSM60UPG
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stainless steel drawer
    Stainless steel drawer
    CA97002-830
    Log in pour la disponibilité
    Each - $999.58
Informations sur le produit
  • Catalog #
    Color
    Material
    Availability
    Price
  • Swivel stem caster, high modulus rubber donut tread
    Swivel stem caster, high modulus rubber donut tread
    CA19766-816
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; Polyolefin hub
    Log in pour la disponibilité
    Each - $137.64
Informations sur le produit
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel/brake stem caster, 300 lbs. load capacity, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Swivel/brake stem caster, 300 lbs. load capacity, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97005-858
    5"
    2.3 lbs.
    Log in pour la disponibilité
    Case of 8 - $657.53
Informations sur le produit
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel/brake stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    Swivel/brake stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    CA97002-900
    5"
    2 lbs.
    Log in pour la disponibilité
    Each - $308.66
Informations sur le produit
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Swivel stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97002-902
    5"
    2.1 lbs.
    Log in pour la disponibilité
    Each - $576.71
Informations sur le produit
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    Swivel stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    CA97002-898
    5"
    2 lbs.
    Log in pour la disponibilité
    Each - $287.29
Informations sur le produit
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Rigid stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Rigid stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97005-860
    5"
    2 lbs.
    Log in pour la disponibilité
    Each - $599.83
Informations sur le produit
  • Catalog #
    Color
    Material
    Availability
    Price
  • Swivel/brake stem caster, high modulus rubber donut tread
    Swivel/brake stem caster, high modulus rubber donut tread
    CA22221-208
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; polyolefin hub
    Log in pour la disponibilité
    Each - $159.49
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stainless steel keyboard tray
    Stainless steel keyboard tray
    CA97002-828
    Log in pour la disponibilité
    Each - $980.89