RTW Adjustable-Height Worktables, Production Basics
About this item
Ergonomic tables are constructed of 12-gauge steel and industrial-grade laminate with an abrasion-resistant finish.
- Optional ESD laminate has static dissipative range of 106 to 109
- Height is adjustable in 5 cm (2") increments from 76.2 to 91.4 cm (30 to 36")
Standard models include blue frame with white laminate work surface or gray ESD laminate work surface.
Load capacity: 750 lbs.
Ordering information: Tables include frame, work surface, four 5×5 cm (2×2") leg adjusters, leveling feet, and parts and hardware packet for assembly. ESD laminate tables also include ground bolt and wire. To order uprights, accessories, or frames and work surfaces in different colors, contact your Avantor representative. Additional frame colors include light neutral, white, gray, and black; laminate colors include gray, almond, ESD beige, and ESD white.
27 Options Available Below
Product Details & Documents
Ergonomic tables are constructed of 12-gauge steel and industrial-grade laminate with an abrasion-resistant finish.
- Optional ESD laminate has static dissipative range of 106 to 109
- Height is adjustable in 5 cm (2") increments from 76.2 to 91.4 cm (30 to 36")
Standard models include blue frame with white laminate work surface or gray ESD laminate work surface.
Load capacity: 750 lbs.
Ordering information: Tables include frame, work surface, four 5×5 cm (2×2") leg adjusters, leveling feet, and parts and hardware packet for assembly. ESD laminate tables also include ground bolt and wire. To order uprights, accessories, or frames and work surfaces in different colors, contact your Avantor representative. Additional frame colors include light neutral, white, gray, and black; laminate colors include gray, almond, ESD beige, and ESD white.
Product Family Options
Tables with Laminate Work Surface
Product Information
- DésignationCapacitéWorksurface Color/FinishWorksurface MaterialWorksurface ThicknessBase/Leg MaterialHauteurW×DShipping WeightAvailabilityPrice
- Adjustable height table750 lbs.White1¼"Painted steel30 - 36"36×30"115 lbs.Log in for availabilityEach$1,187.42Supplier Number: 3001
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"48×30"120 lbs.Log in for availabilityEach$1,277.08Supplier Number: 3000
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"60×30"130 lbs.Log in for availabilityEach$1,241.96Supplier Number: 3005
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"72×30"140 lbs.Log in for availabilityEach$1,352.70Supplier Number: 3010
- Adjustable height table, 72"frame750 lbs.WhiteLaminate1¼"Painted steel30 - 36"96×36"165 lbs.Log in for availabilityEach$1,590.78Supplier Number: 3014
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"48×36"125 lbs.Log in for availabilityEach$1,283.06Supplier Number: 3002
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"60×36"135 lbs.Log in for availabilityEach$1,412.10Supplier Number: 3007
- Adjustable height table750 lbs.WhiteLaminate1¼"Painted steel30 - 36"72×36"145 lbs.Log in for availabilityEach$1,519.65Supplier Number: 3011
- Adjustable height table, 72"frame750 lbs.WhiteLaminate1¼"Painted steel30 - 36"96×36"175 lbs.Log in for availabilityEach$1,573.11Supplier Number: 3015
Tables with ESD Laminate Work Surface
Product Information
- DésignationCapacitéWorksurface Color/FinishWorksurface MaterialWorksurface ThicknessBase/Leg MaterialHauteurW×DShipping WeightAvailabilityPrice
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"36×30"115 lbs.Log in for availabilityEach$1,609.87Supplier Number: 3109
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"48×30"120 lbs.Log in for availabilityEach$1,590.22Supplier Number: 3100
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"60×30"130 lbs.Log in for availabilityEach$1,806.39Supplier Number: 3105
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"72×30"140 lbs.Log in for availabilityEach$1,996.72Supplier Number: 3110
- Adjustable height ESD table, 72"frame750 lbs.Gray1¼"Painted steel30 - 36"96×36"165 lbs.Log in for availabilityEach$2,318.14Supplier Number: 3114
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"36×36"120 lbs.Log in for availabilityEach$1,713.42Supplier Number: 3103
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"48×36"125 lbs.Log in for availabilityEach$1,685.05Supplier Number: 3102
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"60×36"135 lbs.Log in for availabilityEach$1,901.27Supplier Number: 3107
- Adjustable height ESD table750 lbs.Gray1¼"Painted steel30 - 36"72×36"145 lbs.Log in for availabilityEach$2,117.68Supplier Number: 3111
- Adjustable height ESD table, 72"frame750 lbs.Gray1¼"Painted steel30 - 36"96×36"175 lbs.Log in for availabilityEach$2,389.13Supplier Number: 3115
Table Frames Only
Product Information
- DésignationCapacitéWorksurface Color/FinishWorksurface MaterialWorksurface ThicknessBase/Leg MaterialHauteurW×DShipping WeightAvailabilityPrice
- Supplier Number: 3063
- RTW frame for 30x48 tabletop750 lbs.Painted steel28¼ - 35¼"48×30"50 lbs.Log in for availabilityEach$820.22Supplier Number: 3050
- RTW frame for 30x60 tabletop750 lbs.Painted steel28¼ - 34¼"60×30"85 lbs.Log in for availabilityEach$846.54Supplier Number: 3055
- RTW frame for 30x72 tabletop750 lbs.Painted steel28¼ - 34¼"72×30"95 lbs.Log in for availabilityEach$894.98Supplier Number: 3060
- Supplier Number: 3030
- RTW frame for 36x48 tabletop750 lbs.Painted steel28¼ - 34¼"48×36"88 lbs.Log in for availabilityEach$848.57Supplier Number: 3052
- RTW frame for 36x60 tabletop750 lbs.Painted steel28¼ - 34¼"60×36"89 lbs.Log in for availabilityEach$914.32Supplier Number: 3057
- RTW frame for 36x72 tabletop750 lbs.Painted steel28¼ - 34¼"72×36"99 lbs.Log in for availabilityEach$941.25Supplier Number: 3061