Protector® Standard Storage Cabinets, Labconco®
Protector® Standard Storage Cabinets, Labconco®
Supplier:   Labconco
Protector® Standard Storage Cabinets, Labconco®
Supplier:  Labconco
Certificates
Protector® Standard Storage Cabinets, Labconco®
Protector® Standard Storage Cabinets, Labconco®
15 Options Available Below
Product Details & Documents
Standard cabinets support Labconco® laboratory hoods and can be used for storage of small laboratory instruments.
  • Available in six widths and two heights. Single-door models can be ordered with left or right hinge opening
Cabinets are constructed of white epoxy-coated steel and include one 20.3 cm (8") filler panel to increase cabinet’s overall depth from 55.9cm (22") to 76.2 cm (30"), for installation flush to wall.
Ordering information: Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Cabinets are supplied with manual-closing, non-locking doors; four leveling feet; and one 20.3 cm (8") filler panel. Shelves (sold separately) are available for all cabinet widths. A work surface (also sold separately) is required for these cabinets, when used to support hoods without integral bottoms.
Product Family Options
Product Information
  • Désignation
    Type
    Largeur
    Hauteur
    Shipping Weight
    REDISHIP
    Availability
    Price
  • Standard base cabinet
    1 door, left hinge
    12"
    35½"
    65 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,650.00
    $2,650.00
    Each
       $2,650.00
    Minimum order quantity is 1
    Supplier Number: 9900800
  • Standard base cabinet
    1 door, left hinge
    12"
    31½"
    60 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,850.00
    $2,850.00
    Each
       $2,850.00
    Supplier Number: 9904500
  • Standard base cabinet
    1 door, right hinge
    12"
    35½"
    65 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,650.00
    $2,650.00
    Each
       $2,650.00
    Minimum order quantity is 1
    Supplier Number: 9900500
  • Standard base cabinet
    1 door, right hinge
    12"
    31½"
    60 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,850.00
    $2,850.00
    Each
       $2,850.00
    Supplier Number: 9904200
  • Standard base cabinet
    1 door, left hinge
    18"
    31½"
    85 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $3,000.00
    $3,000.00
    Each
       $3,000.00
    Supplier Number: 9904400
  • Standard base cabinet
    1 door, right hinge
    18"
    35½"
    90 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,800.00
    $2,800.00
    Each
       $2,800.00
    Minimum order quantity is 1
    Supplier Number: 9900400
  • Standard base cabinet
    1 door, right hinge
    18"
    31½"
    85 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $3,000.00
    $3,000.00
    Each
       $3,000.00
    Supplier Number: 9904100
  • Standard base cabinet
    1 door, left hinge
    24"
    35½"
    130 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,920.00
    $2,920.00
    Each
       $2,920.00
    Minimum order quantity is 1
    Supplier Number: 9900600
  • Standard base cabinet
    1 door, left hinge
    24"
    31½"
    120 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $3,100.00
    $3,100.00
    Each
       $3,100.00
    Supplier Number: 9904300
  • Standard base cabinet
    1 door, right hinge
    24"
    35½"
    130 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,920.00
    $2,920.00
    Each
       $2,920.00
    Minimum order quantity is 1
    Supplier Number: 9900300
  • Standard base cabinet
    1 door, right hinge
    24"
    31½"
    120 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $3,100.00
    $3,100.00
    Each
       $3,100.00
    Supplier Number: 9904000
  • Standard base cabinet (for precise glove boxes)
    2 doors
    30"
    35½"
    155 lbs.
    Yes
    Log in for availability
    Log in for availability
    Each
    Each - $3,060.00
    $3,060.00
    Each
       $3,060.00
    Supplier Number: 9900200
  • Standard base cabinet
    2 doors
    36"
    35½"
    180 lbs.
    Yes
    Log in for availability
    Log in for availability
    Each
    Each - $3,150.00
    $3,150.00
    Each
       $3,150.00
    Supplier Number: 9900100
  • Standard base cabinet
    2 doors
    48"
    35½"
    260 lbs.
    Yes
    Log in for availability
    Log in for availability
    Each
    Each - $3,440.00
    $3,440.00
    Each
       $3,440.00
    Supplier Number: 9900000
  • Standard base cabinet
    1 door, left hinge
    18"
    35½"
    90 lbs.
    No
    Log in for availability
    Log in for availability
    Each
    Each - $2,800.00
    $2,800.00
    Each
       $2,800.00
    Minimum order quantity is 1
    Supplier Number: 9900700
Accessories
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-120
    12"
    84"
    Log in for availability
    Each - $384.00
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-122
    18"
    72"
    Log in for availability
    Each - $399.00
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-124
    24"
    72"
    Log in for availability
    Each - $420.00
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-126
    30"
    67"
    Log in for availability
    Each - $441.00
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-128
    36"
    65"
    Log in for availability
    Each - $455.00
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Shelf kit
    Shelf kit
    46620-130
    48"
    67"
    Log in for availability
    Each - $476.00
Product Information
  • Catalog #
    Height
    Shipping Weight
    Availability
    Price
  • Shelf kit
    Shelf kit
    470203-302
    65"
    15 lbs.
    Log in for availability
    Each - $476.00
Product Information
  • Catalog #
    Height
    Shipping Weight
    Availability
    Price
  • Shelf kit
    Shelf kit
    470203-304
    37"
    12 lbs.
    Log in for availability
    Each - $455.00
Product Information
  • Catalog #
    Height
    Shipping Weight
    Availability
    Price
  • Shelf kit
    Shelf kit
    470203-306
    35"
    10 lbs.
    Log in for availability
    Each - $441.00
Product Information
  • Catalog #
    Height
    Shipping Weight
    Availability
    Price
  • Shelf kit
    Shelf kit
    470203-308
    46"
    8 lbs.
    Log in for availability
    Each - $420.00