Solid Epoxy Flat Work Surfaces, Labconco®
Supplier: Labconco
Jump to:
À propos de cet article
Combine with a base stand or cabinet to support Fiberglass 30 and Basic™ Labconco® laboratory hoods.
- For Use with Base Stands and Cabinets
- Designed to Support Fiberglass 30 and Basic Labconco Laboratory Hoods
- Black Surfaces
- Depth: 76.2 cm (30")
- Thickness: 3.2 cm (1¼")
Surfaces are black.
4 Options Available Below
Product Details & Documents
Combine with a base stand or cabinet to support Fiberglass 30 and Basic™ Labconco® laboratory hoods.
- For Use with Base Stands and Cabinets
- Designed to Support Fiberglass 30 and Basic Labconco Laboratory Hoods
- Black Surfaces
- Depth: 76.2 cm (30")
- Thickness: 3.2 cm (1¼")
Surfaces are black.
Product Family Options
Informations sur le produit
- DésignationFor Use WithWidthShipping WeightAvailabilityPrice
- Solid Epoxy Flat Work Surface36" Wide Base Cabinets or Stands3 ft.90 lbs.Log in pour la disponibilitéEach$2,350.00Supplier Number: 4882805
- Supplier Number: 4882803
- Supplier Number: 4882804
- Solid Epoxy Flat Work Surface30" Wide Base Cabinets2½ ft.86 lbs.Log in pour la disponibilitéEach$2,080.00Supplier Number: 4882806