Electrical Receptacle Kits for Basic™ Laboratory Hoods, Labconco®
Supplier:   Labconco
Electrical Receptacle Kits for Basic™ Laboratory Hoods, Labconco®
Supplier:  Labconco
Certificates
Electrical Receptacle Kits for Basic™ Laboratory Hoods, Labconco®
Electrical Receptacle Kits for Basic™ Laboratory Hoods, Labconco®
2 Options Available Below
Product Details & Documents
Kits include electrical receptacle, junction box, receptacle cover plate, and installation instructions
Electrical wiring not included.
Product Family Options
Informations sur le produit
  • Désignation
    Electrical
    Availability
    Price
  • Standard Duplex Electrical Receptacle Kit
    115 V, 20 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $70.70
    $70.70
    Each
       $70.70
    Supplier Number: 4844400
  • GFI Duplex Electrical Receptacle Kit
    115 V, 20 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.00
    $171.00
    Each
       $171.00
    Supplier Number: 4844401