VWR® Circulus™ Magnetic Stir Bars, Color-Coded | Avantor
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
Supplier:  VWR International
Certificates
VWR® Circulus™  Magnetic Stir Bars, Color-Coded
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
1 Options Available Below
Product Details & Documents
These magnetic stir bars perform well with all problem liquids, highly viscous mixtures, metal particles in fluids, and round (convex) bottom vessels.
  • Excellent self-centering characteristics
  • Fast mixing action with high turbulence
  • Greater stability
  • Less drifting at higher speeds
Available in two lengths, 36 and 54 mm (1 7/16 and 2 1/8"), each is designed as a 8 mm (0.315”) diameter bar with solid, color-coded, 20 mm (0.79") diameter, Teflon® resin end discs.
Product Family Options
Informations sur le produit
  • Longueur
    Ø
    Revêtement
    Color
    Availability
    Price
  • 36 mm (1⁷⁄₁₆")
    7.9 mm (5/16 ")
    PTFE
    White
    Discontinued
  • 36 mm (1⁷⁄₁₆")
    7.9 mm (5/16 ")
    PTFE
    Red
    Discontinued
  • 36 mm (1⁷⁄₁₆")
    7.9 mm (5/16 ")
    PTFE
    Yellow
    Discontinued
  • 36 mm (1⁷⁄₁₆")
    7.9 mm (5/16 ")
    PTFE
    Blue
    Discontinued
  • 54 mm (2⅛")
    7.9 mm (5/16 ")
    PTFE
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $38.21
    $38.21
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $1,910.38
    $1,910.38
    Prix promotionnel pour les nouveaux clients
    Each
       $38.21
    Case of 50
       $1,910.38
  • 54 mm (2⅛")
    7.9 mm (5/16 ")
    PTFE
    Red
    Discontinued
  • 54 mm (2⅛")
    7.9 mm (5/16 ")
    PTFE
    Yellow
    Discontinued
  • 54 mm (2⅛")
    7.9 mm (5/16 ")
    PTFE
    Blue
    Discontinued