Synthware Separatory Funnel with PTFE Stopcock, Kemtech America
Funnels are available with a glass or PTFE stopper.
- Heavy wall borosilicate construction
- Ground glass joint
- Teflon® or glass(A) stopper
- Teflon® stopcock
Designed for efficient separation of immiscible liquids.
Frequently Bought Together