About this item
The NovaTaq™ PCR Master Mix is a ready-to-use mixture of ultrapure deoxynucleotides
Offers advantages of time-savings, consistency, and minimal risk of contamination. Supplied in package sizes of 100rxn (100µL/rxn).
Components: 2X concentrated NovaTaq DNA polymerase, Ultrapure deoxynucleotides, Reaction buffer without MgCl2, 4x1.25 mL for 2x Nova Taq PCR Master Mix, 1.5 mL for 25 mM MgCl2 solution, 6 mL for PCR grade water.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.