VWR® Incubating Microplate Shaker | Avantor
VWR® Incubating Microplate Shaker
   Options durables disponibles
Supplier:   VWR International
VWR® Incubating Microplate Shaker
Supplier:  VWR International
   Options durables disponibles
Certificates
VWR® Incubating Microplate Shaker
1 Options Available Below
Product Details & Documents
Incubating microplate shakers are ideal for cell cultures, hybridizations, and suspensions.
  • Optimized for shaking microplates or microtubes
  • Digital display for speed, temperature, and time
  • Microprocessor control
  • Timer with audible alarm
  • Available with opaque lid for light-sensitive samples
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor PID temperature controller maintains precise temperature control from 5° above ambient to 65 °C (149 °F)
The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. A Lexan® polycarbonate lid permits viewing of samples from all sides without disturbing internal temperature. Platform dimensions: 19.7Wx27.9Dcm (73/4x11").
Optional microtube rack attaches to platform to hold 1.5 ml or 2 ml microtubes and has a capacity of 70 tubes. Units with opaque lid are designed for light sensitive samples.
Easy-to-use controls allow user to adjust temperatures in 1 °C increments. Built-in timer with LED display will show elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. Holds four microplates or two microtube racks.
Certifications: RoHS certified. TUV Listed, CE Marked. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 2.3 m (7.5') detachable, three-wire cord and plug. Supplied without accessories which must be ordered separately. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 12620-930
  • 97043-606
  • 10027-128
  • 10027-132
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Temperature range:
  • Ambient +5…65 °C (149 °F)
  • Ambient +5…65 °C (149 °F)
  • Ambient +5…65 °C (149 °F)
  • Ambient +5…65 °C (149 °F)
  • Temperature uniformity:
  • ±0.5°C at 37°C
  • ±0.5°C at 37°C
  • ±0.5°C at 37°C
  • ±0.5°C at 37°C
  • Speed accuracy:
  • ±2%
  • ±2%
  • ±2%
  • ±2%
  • Max. load:
  • 2.3 kg (5 lbs.)
  • 2.3 kg (5 lbs.)
  • 2.3 kg (5 lbs.)
  • 2.3 kg (5 lbs.)
  • Overall dimensions:
  • 28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
  • 28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
  • 28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
  • 28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
  • Timer:
  • 1 second to 160 hours
  • 1 second to 160 hours
  • 1 second to 160 hours
  • 1 second to 160 hours
  • Shipping weight:
  • 13.6 kg (30 lbs.)
  • 13.6 kg (30 lbs.)
  • 13.6 kg (30 lbs.)
  • 13.6 kg (30 lbs.)
Spécifications
Product Family Options
Shakers
Informations sur le produit
  • Désignation
    Electrical
    Orbit
    Speed Range
    Plage de température
    Availability
    Price
  • Incubating Microplate Shaker
    120 V, 5 A, 450 W
    3 mm (⅛")
    100 - 1200 rpm
    5° above ambient...65 °C (149 °F)
    Discontinued
    Spécifications:
    CSA Certified
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    12620-930
    Environmentally Preferable
    Waste Reducing
    Temperature range
    Ambient +5…65 °C (149 °F)
    Temperature uniformity
    ±0.5°C at 37°C
    Speed accuracy
    ±2%
    Max. load
    2.3 kg (5 lbs.)
    Overall dimensions
    28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
    Timer
    1 second to 160 hours
    Shipping weight
    13.6 kg (30 lbs.)
    CSA Certified
    Product available on GSA Advantage®
      Waste Reducing
  • Incubating Microplate Shaker with Opaque Lid
    120 V, 5 A, 450 W
    3 mm (⅛")
    100 - 1200 rpm
    5° above ambient...65 °C (149 °F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,023.48
    $8,023.48
    Each
       $8,023.48
    Spécifications:
    CSA Certified
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    97043-606
    Environmentally Preferable
    Temperature range
    Ambient +5…65 °C (149 °F)
    Temperature uniformity
    ±0.5°C at 37°C
    Speed accuracy
    ±2%
    Max. load
    2.3 kg (5 lbs.)
    Overall dimensions
    28W x 19.7H x 43.2D cm (11 x 73/4 x 17")
    Timer
    1 second to 160 hours
    Shipping weight
    13.6 kg (30 lbs.)
    CSA Certified
    Product available on GSA Advantage®
Accessories
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Half rack for 70×1.5 - 2 ml microtubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $364.01
    $364.01
    Each
       $364.01