Synthware Stopcock, Glass Plug, T-Bore
Fournisseur: Kemtech America
Certificats
À propos de cet article
Stopcock with solid glass T-bore plug.
- T-bore
- Heavy wall borosilicate construction
- All glass stopcock
Supplied with stainless steel constant pressure retention spring and clip.
Frequently Bought Together