Synthware Stopcock, 1:5 PTFE Plug, Double Oblique Bore, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Stopcocks have 1:5 taper PTFE plug with double oblique bore.
- Teflon® stopcock construction
- Double Bblique bore
- Heavy wall corosilicate Construction
- Serrated hose connections
Stopcock barrels are polished to prevent scratches.
Frequently Bought Together