Synthware Stopcock, 1:5 PTFE Plug, Metering Valve, Kemtech America
Catalog # 60005-126
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Plug
- Material:PTFE
- Environmentally Preferable:
- Cat. no.:60005-126
- Stopcock Size:4 mm
- Supplier No.:S191530
Specifications
About this item
Replacement Teflon® metering stopcock.
- Teflon® construction
- Metering valve
- Size 15
With a threaded Teflon® needle valve for the fine control of flowthrough the bore of the stopcock plug. This replacement kit is complete with all the components.