Synthware Stopcock, 1:5 PTFE Plug, Straight Bore, Kemtech America
Stopcocks have a 1:5 taper PTFE plug and a threaded pressure nut
- Straight Bore
- Heavy wall borosilicate construction
- Serrated hose connections
Stopcock barrels are polished to prevent scratches.
Frequently Bought Together