Synthware Separatory Funnels
Supplier: Kemtech America
Jump to:
À propos de cet article
Funnels are equipped with a 2 mm PTFE stopcock.
- Heavy wall Borosilicate construction
- Ground glass joints
- Teflon® stopcock
- Available with Glass (A) or Teflon® Stopper
Available with a glass or PTFE stopper.
24 Options Available Below
Product Details & Documents
Funnels are equipped with a 2 mm PTFE stopcock.
- Heavy wall Borosilicate construction
- Ground glass joints
- Teflon® stopcock
- Available with Glass (A) or Teflon® Stopper
Available with a glass or PTFE stopper.
Product Family Options
Informations sur le produit
- DésignationCapacitéST Joint SizeStopcock DiameterStopper MaterialAvailabilityPrice
- Supplier Number: F451010
- Supplier Number: F451010A
- Supplier Number: F451025
- Supplier Number: F459025
- Supplier Number: F451025A
- Supplier Number: F451050
- Supplier Number: F459050
- Supplier Number: F451050A
- Supplier Number: F451100
- Supplier Number: F459100
- Supplier Number: F451100A
- Supplier Number: F459100A
- Supplier Number: F451125
- Supplier Number: F459125
- Supplier Number: F454125
- Supplier Number: F451125A
- Supplier Number: F459125A
- Supplier Number: F454125A
- Supplier Number: F451250
- Supplier Number: F459250
- Supplier Number: F454250
- Supplier Number: F451250A
- Supplier Number: F459250A
- Supplier Number: F454250A
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm glass plug
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 4 mm straight-bore glass plug only
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm PTFE Plug
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE plug, 4 mm