Synthware Flasks with High Vacuum Valve
Synthware Flasks with High Vacuum Valve
Supplier:   Kemtech America
Synthware Flasks with High Vacuum Valve
Supplier:  Kemtech America
Certificates
Synthware Flasks with High Vacuum Valve
Synthware Flasks with High Vacuum Valve
7 Options Available Below
Product Details & Documents
Flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • Hi Vacuum Valves
  • Externally threaded
  • Heavy wall borosilicate construction
  • Ground glass joints
Our externally threaded storage flask is designed for storage of air sensitive materials.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Valve Size
    Availability
    Price
  • 10 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $186.78
    $186.78
    Prix promotionnel pour les nouveaux clients
    Each
       $186.78
    Minimum order quantity is 1
    Supplier Number: F490010
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-232
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 50 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $221.84
    $221.84
    Prix promotionnel pour les nouveaux clients
    Each
       $221.84
    Minimum order quantity is 1
    Supplier Number: F490050
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-236
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 100 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $233.59
    $233.59
    Prix promotionnel pour les nouveaux clients
    Each
       $233.59
    Minimum order quantity is 1
    Supplier Number: F490100
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-238
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 250 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.25
    $210.25
    Prix promotionnel pour les nouveaux clients
    Each
       $210.25
    Minimum order quantity is 1
    Supplier Number: F490250
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-240
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 250 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $275.34
    $275.34
    Prix promotionnel pour les nouveaux clients
    Each
       $275.34
    Minimum order quantity is 1
    Supplier Number: F490258
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-242
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 500 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $222.24
    $222.24
    Prix promotionnel pour les nouveaux clients
    Each
       $222.24
    Minimum order quantity is 1
    Supplier Number: F490500
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-244
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • 1000 ml
    Synthware flask with high vacuum valve
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $271.14
    $271.14
    Prix promotionnel pour les nouveaux clients
    Each
       $271.14
    Minimum order quantity is 1
    Supplier Number: F49081L
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    60003-250
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Accessories
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring
    Viton® O-Ring
    60005-004
    9.2×2.62 mm
    110
    Log in pour la disponibilité
    Each - $4.74
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Plug and control knob for 0 - 12 mm valve
    Plug and control knob for 0 - 12 mm valve
    60004-018
    Log in pour la disponibilité
    Each - $131.50
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring
    Viton® O-Ring
    60004-994
    6.07×1.78 mm
    010
    Log in pour la disponibilité
    Each - $2.67