Synthware Storage Tube with Outer Joint, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Storage tubes are equipped with a high vacuum valve that features a PTFE O-ring at the tip.
- Hi Vacuum valves
- Externally Threaded
- Heavy wall Borosilicate construction
- Ground glass joints
Our externally threaded storage tube is designed for storage of air sensitive materials.
Frequently Bought Together