Synthware Storage Tubes with High Vacuum Valve, Kemtech America
Storage tubes are equipped with a high vacuum valve that features a PTFE O-ring at the tip, and a PTFE wiper to protect the Viton® O-ring on the shaft.
- Hi vacuum valves
- Internally threaded
- Heavy wall borosilicate construction
- Ground glass joints
Our internally threaded storage tube is designed for storage of air sensitive materials.
Frequently Bought Together