PYREX® Square Bottles, Graduated, Corning
PYREX® Square Bottles, Graduated, Corning
Supplier:   Corning
PYREX® Square Bottles, Graduated, Corning
Supplier:  Corning
Certificates
PYREX® Square Bottles, Graduated, Corning
PYREX® Square Bottles, Graduated, Corning
5 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass, with linerless, one-piece, orange PP screw cap with a drip-free pouring ring.
  • Exceptional chemical and thermal resistance
  • Rounded edges allow easy cleaning
  • White graduations and a large marking spot
  • Glass ridge moulded into the bottle indicates full capacity
  • Autoclavable
Square design saves 13 to 20% of shelf space, and slightly convex walls minimize glass-to-glass contact during autoclaving.
Further PP colored screw caps for ease of sorting and identification, vented membrane screw caps with 0.22 µm PTFE hydrophobic membrane, PBT solid or open top screw caps and ETFE pouring rings are optionally available.
Caution: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Square bottles should not be used with bottle top filter units, or in other applications involving vacuum pressure as breakage may occur.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Taille du bouchon
    Plage de graduation
    Intervalle de graduation
    Availability
    Price
  • 100 ml (3.4 oz.)
    Square Bottle
    GL32
    40–80 mL
    20 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.40
    $26.40
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $264.00
    $264.00
    Each
       $26.40
    Case of 10
       $264.00
    Supplier Number: 1396-100
  • 250 ml (8.5 oz.)
    Square Bottle
    GL45
    50–200 mL
    50 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $392.25
    $392.25
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $392.25
    Supplier Number: 1396-250
  • 500 ml (16.9 oz.)
    Square Bottle
    GL45
    100–400 mL
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $345.92
    $345.92
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $345.92
    Supplier Number: 1396-500
  • 1 L (33.8 oz.)
    Square Bottle
    GL45
    100–900 mL
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $451.20
    $451.20
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $451.20
    Supplier Number: 1396-1L
  • 100 ml (3.4 oz.)
    Square Bottle
    GL32
    40–80 mL
    20 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $264.00
    $264.00
    Case of 10
       $264.00
    Supplier Number: 1396-100
Accessories
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • High-Temperature PBT Cap, Red, Open
    High-Temperature PBT Cap, Red, Open
    11310-688
    GL45
    Log in pour la disponibilité
    Case of 10 - $191.00
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • PTFE-Faced Silicone Septa
    PTFE-Faced Silicone Septa
    11310-704
    GL25
    Log in pour la disponibilité
    Case of 10 - $57.10
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • Silicone Septa
    Silicone Septa
    11310-694
    GL45
    Log in pour la disponibilité
    Case of 10 - $163.00
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • High-Temperature PBT Cap, Red, Open
    High-Temperature PBT Cap, Red, Open
    11310-692
    GL25
    Log in pour la disponibilité
    Case of 10 - $81.26
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • PTFE-Faced Silicone Septa
    PTFE-Faced Silicone Septa
    11310-700
    GL45
    Log in pour la disponibilité
    Case of 10 - $137.00
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • PTFE-Faced Silicone Septa
    PTFE-Faced Silicone Septa
    11310-702
    GL32
    Log in pour la disponibilité
    Case of 10 - $77.20
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • High-Temperature PBT Cap, Red, Open
    High-Temperature PBT Cap, Red, Open
    11310-690
    GL32
    Log in pour la disponibilité
    Case of 10 - $143.00
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • Silicone Septa
    Silicone Septa
    11310-698
    GL25
    Log in pour la disponibilité
    Case of 10 - $49.80
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • Silicone Septa
    Silicone Septa
    11310-696
    GL32
    Log in pour la disponibilité
    Case of 10 - $46.07