[ST] Joint Clips
[ST] Joint Clips
Supplier:   Kemtech America
[ST] Joint Clips
Supplier:  Kemtech America
Certificates
[ST] Joint Clips
[ST] Joint Clips
5 Options Available Below
Product Details & Documents
Made of plastic.
  • Heavy duty plastic construction
  • Color coded
  • Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Product Family Options
Product Information
  • Désignation
    ST Joint Size
    Availability
    Price
  • Plastic joint clip
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $11.59
    $11.59
    New Customer Promotional Price
    Each
       $11.59
    Minimum order quantity is 1
    Supplier Number: CS001420
  • Plastic joint clip
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $11.74
    $11.74
    New Customer Promotional Price
    Each
       $11.74
    Minimum order quantity is 1
    Supplier Number: CS001922
  • Plastic joint clip
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $10.07
    $10.07
    New Customer Promotional Price
    Each
       $10.07
    Supplier Number: CS002440
  • Plastic joint clip
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $13.22
    $13.22
    New Customer Promotional Price
    Each
       $13.22
    Minimum order quantity is 1
    Supplier Number: CS002942
  • Plastic joint clip
    34/45
    Log in for availability
    Log in for availability
    Each
    Each - $11.86
    $11.86
    New Customer Promotional Price
    Each
       $11.86
    Minimum order quantity is 1
    Supplier Number: CS003445