VWR® Talon® Ultra Flex Support Systems with Bench Clamp
À propos de cet article
Bench clamp easily attaches to the side of a benchtop or countertop to utilize more space in the lab.
- Unique, flexible arm
- Provided with vinyl and fiberglass sleeves
- Fiberglass sleeves are suited to higher-temperature applications
- Options suited to different-shaped apparatuses
Systems are extremely versatile with a unique, flexible arm that can be placed in virtually any position or angle
The spring clamp head features locking wing nuts that apply tension to the nickel-plated jaws. It is designed to hold thermometers, glass tubes, and temperature probes.
The two- and three-prong clamps are constructed of cast alloy with dual adjust for greater flexibility. Both clamps are provided with vinyl and fiberglass sleeves. The fiberglass sleeves are recommended for higher temperature applications up to 100°C (212°F). The two-prong clamp is ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks. The three-prong clamp is ideal for holding irregular-shaped objects.
Systems include a two-prong clamp, a three-prong clamp, a spring clamp head, a nickel-plated flex arm of either 30.5 cm (12") or 45.7 cm (18") length, an aluminum bench clamp, and an utility wrench.
2 Options Available Below
Product Details & Documents
Bench clamp easily attaches to the side of a benchtop or countertop to utilize more space in the lab.
- Unique, flexible arm
- Provided with vinyl and fiberglass sleeves
- Fiberglass sleeves are suited to higher-temperature applications
- Options suited to different-shaped apparatuses
Systems are extremely versatile with a unique, flexible arm that can be placed in virtually any position or angle
The spring clamp head features locking wing nuts that apply tension to the nickel-plated jaws. It is designed to hold thermometers, glass tubes, and temperature probes.
The two- and three-prong clamps are constructed of cast alloy with dual adjust for greater flexibility. Both clamps are provided with vinyl and fiberglass sleeves. The fiberglass sleeves are recommended for higher temperature applications up to 100°C (212°F). The two-prong clamp is ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks. The three-prong clamp is ideal for holding irregular-shaped objects.
Systems include a two-prong clamp, a three-prong clamp, a spring clamp head, a nickel-plated flex arm of either 30.5 cm (12") or 45.7 cm (18") length, an aluminum bench clamp, and an utility wrench.
Specifications for Product Family
Spécifications
- Catalog No:
- 12621-212
- 12621-214
- Arm Ø:
- 1.3 cm (1/2")
- 1.3 cm (1/2")
Spécifications
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.12621-212Arm Ø1.3 cm (1/2")Product available on GSA Advantage® - Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.12621-214Arm Ø1.3 cm (1/2")Product available on GSA Advantage®
Accessories
Informations sur le produit
- Catalog #Maximum Grip SizeAvailabilityPrice
- Three-Prong Clamp
Informations sur le produit
- Catalog #Maximum Grip SizeAvailabilityPrice
- Bench Clamp
Informations sur le produit
- Catalog #Maximum Grip SizeAvailabilityPrice
- Spring Clamp Head
Informations sur le produit
- Catalog #AvailabilityPrice
- Flexible Arm, 45.7 cm (18")
Informations sur le produit
- Catalog #AvailabilityPrice
- Flexible Arm, 30.5 cm (12")
Informations sur le produit
- Catalog #Maximum Grip SizeAvailabilityPrice
- Two-Prong Clamp