Prefilter for Masks, 3M™
Prefilter for Masks, 3M™
Supplier:   3M
Prefilter for Masks, 3M™
Supplier:  3M
Certificates
Prefilter for Masks, 3M™
Prefilter for Masks, 3M™
4 Options Available Below
Product Details & Documents
For use with 5000-series respirators.
  • NIOSH 42 CFR 84 approved
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Product Family Options
Informations sur le produit
  • Désignation
    Approved For Use Against
    Availability
    Price
  • P95 particulate filter "solids from processing minerals, coal, iron ore, cotton, flour; metal fumes from welding, brazing, cutting of metals; liquid/oil based particles from sprays"
    Protects against solids from processing minerals, coal, iron ore, cotton, flour, metal fumes from welding, brazing, cutting of metals, liquid/oil based particles from sprays that do not emit harmful vapors, use with 5000 series respirators
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $8.60
    $8.60
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $430.00
    $430.00
    Pack of 2
       $8.60
    Case of 100
       $430.00
    Supplier Number: 7000002058
  • N95 filter for use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826
    For use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826
    Discontinued
    Supplier Number: -
  • P95 prefilter "oil and non-oil environments" for use with 5000 and 6000 series respirators
    For oil and non-oil environments, use with 5000 and 6000 series respirators
    Discontinued
    Supplier Number: -
  • Prefilter retainer for use with 56221-054 prefilter.
    Discontinued
    Supplier Number: -
  • Prefilter adapter for use with 56222-931 filter.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $105.00
    $105.00
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $525.00
    $525.00
    Pack of 20
       $105.00
    Case of 100
       $525.00
    Supplier Number: 7000001931
  • P100 filter "solid and liquid particles including those containing oil"
    Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $13.10
    $13.10
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $655.00
    $655.00
    Pack of 2
       $13.10
    Case of 100
       $655.00
    Supplier Number: 7100242461
  • P100 filter "nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluorid, solid and liquid particles including oil" for use with 3M™ 5000, 6000, and 7000 series respirators
    Provides relief from nuisance odors generated from chlorine, sulfur dioxide and hydrogen fluoride, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $18.20
    $18.20
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $910.00
    $910.00
    Pack of 2
       $18.20
    Case of 100
       $910.00
    Supplier Number: 7000002048
  • P100 filter "organic vapor odors, solid and liquid particles including oil" for use with 3M™ 5000, 6000, 7000 series respirators
    Removes organic vapor odors below the OSHA PELs, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, 7000 series respirators
    Discontinued
    Supplier Number: -
  • P95 filter "organic vapor and acid gas odors"
    Provides relief from nuisance level organic vapor and acid gas odors below the OSHA PELs
    Discontinued
    Supplier Number: -