E-LAB 2 Vacuum Pump, Edwards | Avantor
E-LAB 2 Vacuum Pump, Edwards
Vacuum Pump
Catalog # CAA65-207-906
Supplier:   Edwards
Fournisseur #:  A65207906
E-LAB 2 Vacuum Pump, Edwards
Catalog # CAA65-207-906
Supplier:  Edwards
E-LAB 2 Vacuum Pump, Edwards
E-LAB 2 Vacuum Pump, Edwards
Vacuum Pump
Price
$15,580.12
Log in pour la disponibilité
Each
Product Details & Documents
This pump was specifically developed for wet chemistry and sample preparation
An internal cartridge design provides easy servicing and the continuous oil recycling permits constant running. The E-LAB 2 pump has a high efficiency mist filter which eliminates any oil smell.
It is ideal for use with rotary evaporators, centrifugal concentrators, vacuum ovens, distillation/extraction apparatus, fume hoods, and vacuum filtration apparatus.
Certifications: CSA certified and CE marked.
Ordering information: Supplied with manufacturer’s one-year warranty. Complete with a standard nozzle kit for connection to any system.
Spécifications
  • Désignation:
    Vacuum Pump