MicroFunnel™ ST Disposable Filter Funnels, Pall® Life Sciences
À propos de cet article
Convenient and economical choice for sterility testing within isolators.
- Economical
- Ideal for aqueous solutions
- Individually bagged
MicroFunnel™ ST filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique
The unique squeeze separation makes membrane retrieval easy. Choose from 0.45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications or 0.2 and 0.45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances. Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
Individually bagged funnels are packaged within an overpack bag. The double bagged design makes them easy to transfer into isolators or more convenient to wipe down for use in cleanrooms and hoods. Chose from a variety of membranes and pore sizes to best suit your testing needs.
: 100mL funnels 40/pkg: packaged 10 individually bagged funnels per overpack bag, 4 overpack bags per box. 300mL funnels 20/pkg: packaged 5 individually bagged funnels per overpack bag, 4 overpack bags per box. Other types of Disposable Filter Funnels are available. See MicroFunnel™ Disposable Filter Funnels for a broad selection of disposable filter funnels, individually bagged for general testing of aqueous solutions. See MicroFunnel™ Plus Disposable Filter Funnels for funnels designed specifically for water system monitoring—sample cup and filter funnel all in one. Sentino™ Microbiology Pump--designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
7 Options Available Below
Product Details & Documents
Convenient and economical choice for sterility testing within isolators.
- Economical
- Ideal for aqueous solutions
- Individually bagged
MicroFunnel™ ST filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique
The unique squeeze separation makes membrane retrieval easy. Choose from 0.45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications or 0.2 and 0.45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances. Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
Individually bagged funnels are packaged within an overpack bag. The double bagged design makes them easy to transfer into isolators or more convenient to wipe down for use in cleanrooms and hoods. Chose from a variety of membranes and pore sizes to best suit your testing needs.
: 100mL funnels 40/pkg: packaged 10 individually bagged funnels per overpack bag, 4 overpack bags per box. 300mL funnels 20/pkg: packaged 5 individually bagged funnels per overpack bag, 4 overpack bags per box. Other types of Disposable Filter Funnels are available. See MicroFunnel™ Disposable Filter Funnels for a broad selection of disposable filter funnels, individually bagged for general testing of aqueous solutions. See MicroFunnel™ Plus Disposable Filter Funnels for funnels designed specifically for water system monitoring—sample cup and filter funnel all in one. Sentino™ Microbiology Pump--designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
Certificates
Recherche par numéro de lotSpecifications for Product Family
Spécifications
- Catalog No:
- CA89047-136
- 89176-162
- CA89176-162
- CA89234-518
- CA28143-562
- CA28143-564
- CA28143-566
- CA28143-568
- CA77391-980
- CA77391-982
- Filtration area:
- 13.46 cm²
- Effective filtration area:
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- 13.46 cm²
- Dimensions:
- 7.6H×6.1W cm (3.0H×2.4"W)
- 8.9H×8.7W cm (3.5H×3.4"W)
- 8.9H×8.7W cm (3.5H×3.4"W)
- 8.9H×8.7W cm (3.5H×3.4"W)
- 7.6H×6.1W cm (3.0H×2.4"W)
- 8.9H×8.7W cm (3.5H×3.4"W)
- 7.6H×6.1W cm (3.0H×2.4"W)
- 8.9H×8.7W cm (3.5H×3.4"W)
- 7.6H×6.1W cm
- 8.9H×8.7W cm (3.5H×3.4"W)
Spécifications
Product Family Options
Informations sur le produit
- DésignationCapacitéTaille de poreAvailabilityPrice
- Supplier Number: 4851Spécifications:
More
Specifications
Cat. No.CA89047-136Filtration areaEffective filtration area13.46 cm²Dimensions7.6H×6.1W cm (3.0H×2.4"W) - Supor® white, gridded membrane300 ml0.2 µmDiscontinued-Supplier Number: 4853Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89176-162Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W)Product available on GSA Advantage® - Supplier Number: 4853Spécifications:
More
Specifications
Cat. No.CA89176-162Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W) - Supplier Number: 4857Spécifications:
More
Specifications
Cat. No.CA89234-518Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W) - Supplier Number: 4750Spécifications:
More
Specifications
Cat. No.CA28143-562Filtration areaEffective filtration area13.46 cm²Dimensions7.6H×6.1W cm (3.0H×2.4"W) - Supplier Number: 4751Spécifications:
More
Specifications
Cat. No.CA28143-564Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W) - GN-6 white, gridded membrane100 ml0.45 µmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA28143-566Filtration areaEffective filtration area13.46 cm²Dimensions7.6H×6.1W cm (3.0H×2.4"W) - GN-6 white, gridded membrane300 ml0.45 µmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA28143-568Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W) - MicroFunnel™ ST filter funnel, white, gridded, Filter media: ME25 mixed cellulose ester100 ml0.45 µmLog in pour la disponibilitéPack of 40505,10 $Supplier Number: 4811MESpécifications:
More
Specifications
Cat. No.CA77391-980Filtration area13.46 cm²Effective filtration area13.46 cm²Dimensions7.6H×6.1W cm - MicroFunnel™ ST filter funnel, white, gridded, Filter media: ME25 mixed cellulose ester300 ml0.45 µmLog in pour la disponibilitéPack of 20331,69 $Supplier Number: 4812MESpécifications:
More
Specifications
Cat. No.CA77391-982Filtration areaEffective filtration area13.46 cm²Dimensions8.9H×8.7W cm (3.5H×3.4"W)
Accessories
Informations sur le produit
- Catalog #DésignationAvailabilityPrice
- Autoclavable Adapters, Clear; For Vacuum Manifold when using Rubber StopperLog in pour la disponibilitéPack of 3161,45 $