Whatman™ PolyVENT™ Integral Vent Filters
Supplier: Cytiva
Certificates
About this item
Whatman PolyVENT disc filters are integral venting filters that work bidirectionally to prevent contamination from entering containers like fermentation vessels during incubation.
- 0.2 µm hydrophobic PTFE air filters are ideal industrial air filter media
- Validated for 50 steam autoclave cycles; compatible with ethylene oxide (EtO)
- Testable by water break through (WBT) test or bubble point testing
- Manufactured in clean room facilities
- Filtration area 500, 1000 and 2000 cm² to support a range of venting applications
Draining or filling of incubators, fermentation tanks, and other vessels requires a venting filter capable of preventing bacterial contamination. With an integral PTFE filter membrane, Whatman PolyVENT acts as an industrial air filter media for sterilization* of gases entering bioreactors such as fermentation tanks.
Can be autoclaved at 121 °C for 20 minutes (maximum 132 °C)
Certifications: Passes USP Class VI biosafety tests for plastics.
Specifications
- Catalog No:
- CA28137-656
- CA28137-653
- CA28137-652
- CA28137-650
- CA28137-658
- Max. pressure:
- 2 bar - forward direction (29 psi - forward direction)
- 2 bar - forward direction (29 psi - forward direction)
- 2 bar - forward direction (29 psi - forward direction)
- 2 bar - forward direction (29 psi - forward direction)
- 2 bar - forward direction (29 psi - forward direction)
- Application:
- Venting (filling).
Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
Electronics (gases). - Venting (filling).
Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
Electronics (gases). - Venting (filling).
Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
Electronics (gases). - Venting (filling).
Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
Electronics (gases). - Venting (filling).
Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
Electronics (gases).
- Filter material:
- Polytetrafluoroethylene (PTFE)
- Polytetrafluoroethylene (PTFE)
- Polytetrafluoroethylene (PTFE)
- Polytetrafluoroethylene (PTFE)
- Polytetrafluoroethylene (PTFE)