Acro® 50 Vent Filters with Emflon® II Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
These 50 mm filters ideal for use in air or gas applications and for venting of bioreactors, fermentation tanks, and carboys.
- Disposable
- Non sterile
- Hydrophobic
- Can be gamma irradiated
Hydrophobic polyvinylidene fluoride (PVDF) Emflon® II 0.2 µm membrane in polypropylene housing. Lightweight device will not crimp tubing. Inlet/outlet hose connections are 6.4 to 12.7 mm (¹/₄ to ¹/₂") stepped hose barbs with slip Luer Int. Ø. in the hose barb. Autoclavable at 121 °C (250 °F) for 20 minutes.
Specifications
- Catalog No:
- CA28143-838
- CA16003-656
- Filtration area:
- 20 cm²
- 20 cm²
- Max. ambient temperature:
- 60 °C (140 °F)
- 60 °C (140 °F)
- Max. operating pressure:
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature