Filter Funnel, 47 mm, Stainless Steel, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
Certificats
À propos de cet article
The 47 mm stainless steel filter funnels are ideal for applications requiring the ultimate in chemical resistance.
- Chemically resistant
- Easy to assemble
- For vacuum filtration
- Overall Length: 22.2 cm (8.7 in.)
- Diameter: 15.2 cm (6.0 in.)
They are quick and easy to assemble; they seal without the need of O-rings.
They are designed for vacuum filtration of biological samples, oils, solvents, etc.
This product is designed for laboratory applications only and is not approved for use in medical, clinical, surgical or other patient applications.
Caution: Note: Not recommended for use with volatile liquids or high purity fuels as explosion may result from the static discharge.
Spécifications
- Catalog No:
- CA28144-958
- CA28144-903
- Effective filtration area:
- 9.6 cm²
- 9.6 cm²
- Sterilisation:
- Provided non-sterile. Autoclavable if desired at 121 - 123 °C (250 - 253 °F) at 1.0 bar (100 kPa, 15 psi) for 15 - 20 min.
- Provided non-sterile. Autoclavable if desired at 121 - 123 °C (250 - 253 °F) at 1.0 bar (100 kPa, 15 psi) for 15 - 20 min.