Unimodule METD Series Workstations, Envirco®
Fournisseur: Envirco
Certificats
À propos de cet article
These self-contained, 100% exhausted, vertical laminar flow workstations are especially suited for operations involving toxic or noxious fumes
Workstations feature stainless steel interior, clear acrylic eyelid, and fluorescent lighting. Stainless steel transition duct allows connection of plenum to customer-supplied, 30.5cm (12") diameter exhaust duct. Work decks lift out for easy cleaning.
HEPA filters (99.99% efficient to 0.3µm) provide a ISO Class 5 (FED-STD-209E Class 100/M3.5) environment per Federal Standard 209. Workstations pull air through a 51% perforated stainless steel grill. Air then passes through a stainless steel tub and out through a stainless steel exhaust plenum. The stainless steel tub has a liquid drain.
Ordering information: Supplied with a two-year limited parts and labor warranty. Includes a three-wire cord with plug. Workstation requires an exhaust system (not included) capable of 110–125% of air supply module cfm.
Frequently Bought Together