Tabletop Horizontal Laminar Flow Workstations, Envirco® | Avantor
Tabletop Horizontal Laminar Flow Workstations, Envirco®
Supplier:   Envirco
Tabletop Horizontal Laminar Flow Workstations, Envirco®
Supplier:  Envirco
Certificates
Tabletop Horizontal Laminar Flow Workstations, Envirco®
Tabletop Horizontal Laminar Flow Workstations, Envirco®
2 Options Available Below
Product Details & Documents
Self-contained multipurpose horizontal laminar flow workstations can be placed on an existing table, open bench, or support stand
The workstation exterior is constructed of welded aluminum with a corrosion-resistant polyurethane enamel finish, and it has removable clear acrylic side panels. The nonglare, plastic laminated work surface is easy to clean and stands 77.5cm (301/2") from the floor. A recessed fluorescent light fixture provides 150 foot-candles of illumination at the work surface.
Workstations utilize a direct-drive continuous-duty 1/3hp motor with sealed-for-life bearings dynamically balanced and mounted on rubber-in-shear vibration isolators. The motor is designed to provide rated airflow through filter loading in excess of 50% over initial static pressure. The motor features solid-state speed control with RFI suppression for adjusting airflow velocity.
Workstations feature HEPA filters that provide an ISO Class 5 (FED-STD-209E Class 100/M3.5) environment. The HEPA filters have a minimum efficiency of 99.99% at 0.3µm or larger. A perforated aluminum diffuser screen protects the HEPA filter while maintaining uniform air velocity within the work area. A 30% ASHRAE washable prefilter is included to extend the life of the HEPA filter.
 : Manufacturer's one-year limited warranty. Workstations include a power cord with three-prong plug. Optional stainless steel work surfaces, lab service fixtures, and duplex electrical receptacles are available; contact your Avantor sales representative for more information.
Product Family Options
Informations sur le produit
  • Désignation
    Electrical
    BTU/Hour
    Width
    Outside Height
    Work Area Dimensions
    Airflow
    Shipping Weight
    Availability
    Price
  • Workstation
    115 V, 7.5 A
    2950
    38"
    33"
    36½×24"
    675 at 90 fpm
    280 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26,772.33
    $26,772.33
    Each
       $26,772.33
    Supplier Number: 10562
  • Workstation
    115 V, 7.5 A
    3000
    50"
    33"
    48½×24"
    900 at 90 fpm
    330 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47,827.29
    $47,827.29
    Each
       $47,827.29
    Supplier Number: 10564
Accessories
Informations sur le produit
  • Catalog #
    Width
    Availability
    Price
  • UV Light
    UV Light
    VF-10603
    36"
    Log in pour la disponibilité
    Each - $6,718.93
Informations sur le produit
  • Catalog #
    Width
    Availability
    Price
  • UV Light,
    UV Light,
    VF-10604
    48"
    Log in pour la disponibilité
    Each - $5,600.05
Informations sur le produit
  • Catalog #
    Width
    Shipping Weight
    Availability
    Price
  • Support Stand
    Support Stand
    VF-10606
    38"
    123.5 lbs.
    Log in pour la disponibilité
    Each - $5,150.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Duplex Electrical Outlet
    Duplex Electrical Outlet
    VF-10607
    Log in pour la disponibilité
    Each - $1,783.83
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Air, Gas, Vacuum Stopcock
    Air, Gas, Vacuum Stopcock
    VF-10611
    Log in pour la disponibilité
    Each - $1,318.96
Informations sur le produit
  • Catalog #
    Width
    Availability
    Price
  • Stainless Steel Workdeck
    Stainless Steel Workdeck
    VF-10612
    36"
    Log in pour la disponibilité
    Each - $5,427.88
Informations sur le produit
  • Catalog #
    Width
    Availability
    Price
  • Stainless Steel Workdeck
    Stainless Steel Workdeck
    VF-10613
    48"
    Log in pour la disponibilité
    Each - $11,554.08
Informations sur le produit
  • Catalog #
    Width
    Shipping Weight
    Availability
    Price
  • Support Stand
    Support Stand
    VF-10631
    50"
    147.7 lbs.
    Log in pour la disponibilité
    Each - $6,563.22