ContactUsLinkComponent
Horizontal Laminar Flow Clean Benches, Envirco®
 :  Envirco
undefined
Horizontal Laminar Flow Clean Benches, Envirco®
 :  Envirco
  • Catalog No:
  • VF-10552
  • VF-10553
  • VF-10555
  • Btu/hour:
  • 3000 for VF-10552, VF-10553, VF-10554; 6000 for VF-10555
  • 3000 for VF-10552, VF-10553, VF-10554; 6000 for VF-10555
  • 3000 for VF-10552, VF-10553, VF-10554; 6000 for VF-10555
  • Electrical:
  • 115V, 7.5A for VF-10552, VF-10553, VF-10554; 115V, 15A for VF-10555
  • 115V, 7.5A for VF-10552, VF-10553, VF-10554; 115V, 15A for VF-10555
  • 115V, 7.5A for VF-10552, VF-10553, VF-10554; 115V, 15A for VF-10555
Frequently Bought Together