KONTES® Dounce Tissue Grinders, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
For cellular work where the nucleus remains intact after homogenization
Borosilicate glass construction. One tube and two pestles are supplied with each tissue grinder. The large clearance pestle is used for initial sample reduction. The small clearance pestle forms the final homogenate.
Frequently Bought Together