ContactUsLinkComponent
20 ml Stainless Steel Micro Container for Waring® Blenders, Eberbach
  58983-208
 :  Eberbach
undefined
20 ml Stainless Steel Micro Container for Waring® Blenders, Eberbach
  58983-208
 :  Eberbach
 :  E8575.00
  • Description:
    20mL Stainless Steel Micro Container for Waring® Blenders, Eberbach
  • Cat. no.:
    58983-208
  • Supplier No.:
    8575
Working capacity 3–20mL
Unit is 17cm (63/4") high. Container: 8.9H x 3.8dia.cm (31/2Hx11/2dia."). Use the included wrench set to disassemble container for cleaning and sterilizing. (Eberbach 8575)
Stainless steel container and blade assembly, with fluoropolymer resin gaskets. Entire unit can be prechilled to provide additional heat absorbing capacity. Fits blender power unit 58984-700 with adapter 58985-225, and other 1 and 2-speed Waring® power units.