POLYTRON® PT 1200 E Handheld Homogenizers, Kinematica
POLYTRON® PT 1200 E Handheld Homogenizers, Kinematica
POLYTRON® PT 1200 E Handheld Homogenizer, 115 V
Catalog # 47751-624
Small Business Enterprise
Supplier:   Kinematica
Supplier #:  11010026
POLYTRON® PT 1200 E Handheld Homogenizers, Kinematica
Catalog # 47751-624
Supplier:  Kinematica
Small Business Enterprise
POLYTRON® PT 1200 E Handheld Homogenizers, Kinematica
POLYTRON® PT 1200 E Handheld Homogenizers, Kinematica
POLYTRON® PT 1200 E Handheld Homogenizer, 115 V
Price
3 946,16 $
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Model PT 1200E features an ergonomic design for repetitive motions and prolonged use, a 100W motor with safety power supply, and fingertip speed control up to 25,000rpm.
  • Efficient sample preparation–because fast dispersion saves time
  • Easy operation with an on-off switch and speed control
  • Lightweight
  • Intelligent variable speed control for long equipment life
  • Low noise
  • Height adjustable stand (available as optional)
  • Switch aggregate quickly with Click & Go coupling
  • Easy aggregate clean-up with EasyClean design
Powerful motor and effective aggregate action combine to rapidly break down plant and animal tissue, heart muscle, and other general homogenizing applications.
For standard dispersing aggregates (generators), the universal geometry uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and it is reliable and efficient. Disposable aggregates are also available.
Ordering information: Dispersing aggregates/generators (required for operation; sold separately) are not included with purchase of homogenizer. Includes power cable and tabletop power supply (1.8 m).
Spécifications
  • Désignation:
    POLYTRON® PT 1200 E Handheld Homogenizer
  • Electrical:
    115 V
  • Cat. No.:
    47751-624
  • Sound level:
    Drive without aggregate:
    ≤ 65 dB(A) at 20,000 rpm;
    ≤ 76 dB(A) at 25,000 rpm
  • Speed range:
    0 to 25,000 rpm
  • Power rating:
    Universal with carbon brushes; 100 W drive power
  • Volume H₂O:
    0,05 - 250 ml, processing vol.
  • Weight:
    Drive: 440 g;
    Power supply: 310 g
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stand for PT 1200E Homogenizers
    Stand for PT 1200E Homogenizers
    47751-634
    Log in pour la disponibilité
    Each - $520.09